Skip to Content
MilliporeSigma

1.04358

Benzonase® ELISA Kit III

for the immunological detection of Benzonase®

Synonym(s):

Endonuclease Detection Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116158
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

The kit should be kept dry, refrigerated at +2°C to +8°C and protected from light

Quality Segment

technique(s)

ELISASandwich: suitable

application(s)

detection
pharma/biopharma processes
pharma/biopharma processes

storage temp.

2-8°C

General description

Benzonase ELISA Kit III was developed for the detection and quantitative determination of Benzonase® endonuclease.
Our SAFC® portfolio of high-quality raw materials for use in biopharmaceutical processing withstands strict quality control procedures plus the documentation and expertise to help our customers meet requirements as defined by the M-Clarity Program.

M-Clarity Program

Our comprehensive portfolio of downstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Ranging from non-GMP grades for low-risk application, to IPEC-PQG GMP for higher-risk applications, we have products covering all your manufacturing needs.

Application

The Benzonase ELISA Kit III was developed for the detection and quantitative determinationof Benzonase® endonuclease in samples from downstream processing where Benzonase® endonuclease is used as a process or purification aid. The assay allows the quantification of the presence of Benzonase® endonuclease down to concentrations of 0.1 ng/mL. Cross-reactions with standards cell culture supernatant are less than 1%. The kit uses purified polyclonal antibodies specific to Benzonase® endonuclease, which have been coated on to polystyrene microtiter plates.

Packaging

1.04358.0001 / 5 x 96 wells plates per kit

1.04358.0002 / 5 x 96 wells plates per kit

Legal Information

Benzonase is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany


pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

10 - Combustible liquids

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library