Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.04178

Sigma-Aldrich

Rhodamine B (C.I.45170)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H31ClN2O3
CAS Number:
Molecular Weight:
479.01
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.21

form

solid

Quality Level

potency

>2000 mg/kg LD50, oral (Rat)

composition

Dye, ≥90% spectrophotometric assay

pH

3-4 (20 °C, 10 g/L in H2O)

mp

199-201 °C

solubility

15 g/L

density

1.31 g/cm3 at 20 °C

bulk density

250 kg/m3

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

InChI

1S/C28H30N2O3.ClH/c1-5-29(6-2)19-13-15-23-25(17-19)33-26-18-20(30(7-3)8-4)14-16-24(26)27(23)21-11-9-10-12-22(21)28(31)32;/h9-18H,5-8H2,1-4H3;1H

InChI key

PYWVYCXTNDRMGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Rhodamine B (C.I. 45170) for microscopy, is a solid dye for fluorescence AFB staining of bacteriological smears and histological tissue sections of human origin. The mechanism is a non-specific binding of fluorescence dye on a substrate, similar to the classical Ziehl-Neelsen method, although the heating of the Rhodamine B color solution can be avoided. The dye has a constant dye content and a reliable high quality, which is one of the major precondition for a reproducible diagnostic staining.

Application

for microscopy

Analysis Note

Dye content (spectrophotometrically): ≥ 90 %
Identity (UV/VIS-Spectrum): passes test
Absorption maximum λ max (ethanol 50 %): 550 - 552 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.003 g/l; ethanol 50 %): 2115 - 2350
TLC-Test: passes test
Loss on drying (110 °C): ≤ 5 %
Suitability for microscopy: passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service