Skip to Content
MilliporeSigma

1.04174

Pararosaniline (chloride) (C.I. 42500)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H18ClN3
CAS Number:
Molecular Weight:
323.82
NACRES:
NA.21
UNSPSC Code:
41116130
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

crystalline

Quality Segment

mp

268-270 °C

solubility

10 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

InChI

1S/C19H17N3.ClH/c20-16-7-1-13(2-8-16)19(14-3-9-17(21)10-4-14)15-5-11-18(22)12-6-15;/h1-12,20H,21-22H2;1H

InChI key

JUQPZRLQQYSMEQ-UHFFFAOYSA-N

General description

Pararosaniline (chloride) (C.I. 42500) for microscopy Certistain®, is a solid dye used to prepare the Schiff’s reagent required for the PAS reaction, for the staining of mucopolysaccharides in histological tissue specimens. The PAS (periodic acid Schiff) reaction is one of the most frequently used chemical methods for histology. As CERTISTAIN® dye it is chemically quality-certified according to highest standards and strict specifications.

Application

for microscopy Certistain®

Legal Information

CERTISTAIN is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Carc. 1B

Storage Class

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library