Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.03762

SAFC

Ammonium iron(III) citrate

about 18% Fe

Synonym(s):

Ammonium iron(III) citrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O7 · xFe · xH3N
CAS Number:
MDL number:
UNSPSC Code:
12352103
EC Index Number:
214-686-6

Quality Level

form

solid

potency

>2000 mg/kg LD50, oral (Rat)

pH

6-8 (20 °C, 100 g/L in H2O)

solubility

1200 g/L

application(s)

pharma/biopharma processes

storage temp.

2-30°C

InChI

1S/C6H8O7.Fe.H3N/c7-3(8)1-6(13,5(11)12)2-4(9)10;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;1H3/q;+3;/p-3

InChI key

FRHBOQMZUOWXQL-UHFFFAOYSA-K

General description

We offer a range of high quality critical raw materials plus the documentation and expertise to help our customers meet stringent regulatory requirements. Our SAFC® portfolio of high-quality products withstands strict quality control procedures.

Application

Ammonium iron(III) citrate is used as an acidic regulator. Our product Ammonium iron(III) citrate about 18% Fe is suitable for use in pharmaceutical processes.

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

also commonly purchased with this product

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service