Skip to Content
MilliporeSigma

1.03750

Yeast extract agar

suitable for microbiology

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.74
UNSPSC Code:
41106212
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

medium granules

Quality Segment

manufacturer/tradename

GranuCult®

pH

6.3-6.7 (25 °C, 35 g/L in H2O, after autoclaving)

solubility

35 g/L

bulk density

750 kg/m3

application(s)

food and beverages
industrial qc, microbiology

storage temp.

15-25°C

suitability

molds, yeasts

Analysis Note

Appearance (clearness): clear
Appearance (colour): yellowish-brown
pH-value (25 °C): 6.3 - 6.7
Growth (Staphylococcus aureus ATCC 25923 (WDCM 00034)): good to very good
Growth (Escherichia coli ATCC 25922 (WDCM 00013)): good to very good
Growth (Candida albicans ATCC 10231 (WDCM 00054)): good to very good
Growth (Geotrichum candidum DSM 1240): good to very good
Growth (Aspergillus brasiliensis (formerly A. niger) ATCC 16404 (WDCM 00053)): good to very good
Growth (Penicillium commune ATCC 10428): good to very good
Growth (Rhodotorula mucilaginosa DSM 70403): good to very good
Incubation: 7 days; 28 °C; aerobic.

Other Notes

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Yeast extract agar


Storage Class

11 - Combustible Solids

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library