Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

1.02994

SAFC

Sorbitol

liquid, (sorbitol syrup, crystallizing), EMPROVE® ESSENTIAL, Ph. Eur., BP, USP, E 420

Pharma Manufacturing

Synonym(s):

D-Sorbitol, Karion, D-Glucitol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14O6
CAS Number:
Molecular Weight:
182.17
Beilstein:
1721899
MDL number:
UNSPSC Code:
12352201
E Number:
E420
Pricing and availability is not currently available.

Agency

BP
Ph. Eur.
USP

Quality Level

vapor density

<1 (vs air)

vapor pressure

<0.1 mmHg ( 25 °C)

product line

EMPROVE® ESSENTIAL

form

liquid
viscous liquid

pH

6 (20 °C in H2O, 50% solution)

mp

98-100 °C (lit.)

density

1.3 g/cm3

application(s)

liquid formulation
pharmaceutical

storage temp.

15-25°C

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)CO

InChI

1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2/t3-,4+,5-,6-/m1/s1

InChI key

FBPFZTCFMRRESA-JGWLITMVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Whether suspensions, emulsions or solutions – oral liquid dosage forms offer a number of advantages: They allow for individual dosing, are easy to swallow and are suitable for pediatric or geriatric applications. On the other hand, formulators face considerable challenges concerning solubility, API stability in aqueous systems, or taste that can often make a decisive difference. Our large product portfolio covers all excipients required for oral liquid formulation, as solvents and surfactants, flavors, ph adjustments and buffers, preservatives and viscosity enhancers.
As part of our EMPROVE® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Sorbitol is a sugar alcohol which is frequently used as sweetener in oral liquid pharmaceutical formulations, as well as bulking agent, sugar substitute or humectant in cosmetics and food and beverage.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service