Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.02839

Millipore

L-Cysteine hydrochloride monohydrate

for biochemistry

Synonym(s):

Cys, L-Cysteinium Chloride Monohydrate, L-Cysteine Hydrochloride, Monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH(NH2)COOH · HCl · H2O
CAS Number:
Molecular Weight:
175.63
Beilstein:
5158059
MDL number:
UNSPSC Code:
12352209
EC Index Number:
200-157-7
NACRES:
NA.21
Pricing and availability is not currently available.

vapor pressure

<0.1 hPa ( 20 °C)

Quality Level

Assay

98.5-101.0% dry basis (alkalimetric)

form

crystalline powder

pH

0.8-1.2 (20 °C, 100 g/L in H2O)

mp

168-170 °C

solubility

650 g/L

density

1.54 g/cm3

bulk density

780 kg/m3

anion traces

sulfate (SO42-): ≤300 ppm

cation traces

Fe: ≤20 ppm
heavy metals (as Pb): ≤10 ppm

storage temp.

2-30°C

SMILES string

O.Cl.N[C@@H](CS)C(O)=O

InChI

1S/C3H7NO2S.ClH.H2O/c4-2(1-7)3(5)6;;/h2,7H,1,4H2,(H,5,6);1H;1H2/t2-;;/m0../s1

InChI key

QIJRTFXNRTXDIP-JIZZDEOASA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Smartphone-based Ellman′s colourimetric methods for the analysis of d-penicillamine formulation and thiolated polymer: Demonstrates the application of L-Cysteine hydrochloride monohydrate in developing colorimetric assays for pharmaceutical analysis, enhancing the accessibility and cost-effectiveness of drug quality control (Phadungcharoen et al., 2019).

  • The effects of the thiolation with thioglycolic acid and l-cysteine on the mucoadhesion properties of the starch-graft-poly(acrylic acid): Focuses on the modification of polymers with L-Cysteine hydrochloride monohydrate to improve their mucoadhesive properties, which is critical for developing more effective drug delivery systems (Gök et al., 2017).

  • On-line pre-reduction of pentavalent arsenicals by thioglycolic acid for speciation analysis by selective hydride generation-cryotrapping-atomic absorption spectrometry: Utilizes L-Cysteine hydrochloride monohydrate in analytical chemistry techniques to improve the detection and measurement of toxic elements in environmental samples, crucial for monitoring and ensuring public health (Musil et al., 2008).

Biochem/physiol Actions

NMDA glutamatergic receptor antagonist.

Analysis Note

Assay (alkalimetric, calculated on dried substance): 98.5 - 101.0 %
Identity (IR-spectrum): passes test
Identity (Chloride): passes test
Appearance: white or almost white, crystalline powder or colorless crystals
Appearance of solution (25 g/l, water): clear and not more intense in color than reference solution BY6
Spec. rotation (α 20/D, 80 g/l, hydrochloric acid 250 g/l, calc. on dried substance): +5.5 to +7.0
Spec. rotation (α 25/D, 80 g/l, hydrochloric acid 6 mol/l, calc. on dried substance): +5.7 to +6.8
Sulfate (SO₄): ≤ 300 ppm
Heavy metals (as Pb): ≤ 10 ppm
Fe (Iron): ≤ 20 ppm
Related Compounds, total (TLC): ≤ 0.5 %
Ninhydrin-positive substances (LC)(impurity A (570 nm)): ≤ 0.5 %
Ninhydrin-positive substances (LC)(any ninhydrin-positive impurity): ≤ 0.2 %
Ninhydrin-positive substances (LC) (ammonium (570 nm)): ≤ 0.02 %
Ninhydrin-positive substances (LC)(total impurities): ≤ 1.0 %
Residual solvents (ICH Q3C): excluded by manufacturing process
Sulfated ash (600 °C): ≤ 0.1 %
Loss on drying (Vacuum < 0.7 kPa, P₂O₅, 24 h): 8.0 - 12.0 %
Bacterial endotoxins: ≤ 6.0 I.U./g

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service