Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.02710

Supelco

Copper(II) acetate monohydrate

greener alternative

cryst. EMPLURA®

Synonym(s):

Copper(II) acetate monohydrate, Cupric acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3COO)2Cu · H2O
CAS Number:
Molecular Weight:
181.63 (anhydrous basis)
MDL number:
UNSPSC Code:
12352103
EC Index Number:
205-553-3
NACRES:
NA.21
form:
solid
solubility:
72 g/L

Quality Level

product line

EMPLURA®

Assay

≥99.0% (iodometric)

form

solid

potency

501 mg/kg LD50, oral (Rat)

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤0.1% In water insoluble matter

pH

5.2-5.5 (20 °C, 20 g/L in H2O)

mp

115 °C

solubility

72 g/L

density

1.88 g/cm3 at 20 °C

bulk density

1100 kg/m3

anion traces

chloride (Cl-): ≤0.01%
sulfate (SO42-): ≤0.2%

cation traces

Fe: ≤0.005%
Zn: ≤0.01%

greener alternative category

storage temp.

2-30°C

SMILES string

[Cu+2].[O-]C(=O)C.[O-]C(=O)C.O

InChI

1S/2C2H4O2.Cu.H2O/c2*1-2(3)4;;/h2*1H3,(H,3,4);;1H2/q;;+2;/p-2

InChI key

NWFNSTOSIVLCJA-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product has been enhanced for catalysis. Click here for more information.

Analysis Note

Assay (iodometric): ≥ 99.0 %
In water insoluble matter: ≤ 0.1 %
pH-value (2 %; water): 5.2 - 5.5
Chloride (Cl): ≤ 0.01 %
Sulfate (SO₄): ≤ 0.2 %
Fe (Iron): ≤ 0.005 %
Zn (Zinc): ≤ 0.01 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service