Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.02561

Sigma-Aldrich

ISOSLIDE Congo Red Control Slides

control slides with reference tissue for the detection of amyloid structures in histological tissue

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116124

Quality Level

application(s)

clinical testing
diagnostic assay manufacturing
hematology

storage temp.

15-25°C

General description

The ISOSLIDE Congo Red Control Slides - with reference tissue for the detection of amyloid structures in histological tissue, are prepared using sections of suitable animal material, which yield good images of amyloid structures in histological tissues. Each package of ISOSLIDE control slides contains 25 slides. The tissue is applied onto the slide next to the white printed labelling field. One slide is already stained with the reference method (Congo Red staining kit, Product number 1.01641) and is used for comparison. The 24 unstained slides are stained according to the corresponding protocol or with the laboratory′s own protocol. After the staining of the control slides, the staining result is to be compared with the laboratory material and the pre-stained control slide.
The ISOSLIDE control slides are not quality-controlled for their application with molecular pathology methods and IHC.This product is registered as IVD and CE marked. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Application

Control slides with reference tissue for the detection of amyloid structures in histological tissue

Legal Information

ISOSLIDE is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service