Skip to Content
MilliporeSigma

1.01984

tert-Butyl methyl ether

for spectroscopy Uvasol®

Synonym(s):

tert-Butyl methyl ether, MTBE, MTB, 2-Methoxy-2-methylpropane, Methyl tert-butyl ether

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(CH3)3COCH3
CAS Number:
Molecular Weight:
88.15
UNSPSC Code:
12191502
EC Index Number:
216-653-1
NACRES:
NA.03
MDL number:
Assay:
≥99.9% (GC)
Technique(s):
UV/Vis spectroscopy: suitable
Bp:
55.3 °C/1013 hPa
Vapor pressure:
268 hPa ( 20 °C)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

268 hPa ( 20 °C)

Quality Segment

assay

≥99.9% (GC)

form

liquid

autoignition temp.

460 °C

potency

>2000 mg/kg LD50, oral (Rat), >2000 mg/kg LD50, skin (Rabbit)

expl. lim.

1.5-8.5 % (v/v)

technique(s)

UV/Vis spectroscopy: suitable

impurities

≤0.0002 meq/g Acidity, ≤0.0002 meq/g Alkalinity, ≤0.01% Water

evapn. residue

≤0.0002%

color

APHA: ≤10

transmittance

215 nm, ≥40%, 235 nm, ≥55%, 240 nm, ≥60%, 255 nm, ≥85%, 260 nm, ≥90%, 280 nm, ≥98%

kinematic viscosity

0.409 cSt(40 °C)

bp

55.3 °C/1013 hPa

mp

-108.6 °C

transition temp

flash point -28 °C

density

0.74 g/cm3 at 20 °C

UV absorption

λ: 215 nm Amax: ≤0.40, λ: 235 nm Amax: ≤0.26, λ: 240 nm Amax: ≤0.22, λ: 255 nm Amax: ≤0.07, λ: 260 nm Amax: ≤0.05, λ: 280 nm Amax: ≤0.01

storage temp.

2-30°C

SMILES string

O(C(C)(C)C)C

InChI

1S/C5H12O/c1-5(2,3)6-4/h1-4H3

InChI key

BZLVMXJERCGZMT-UHFFFAOYSA-N

General description

Accurate analytic results in UV/VIS and infrared spectroscopy depend on the use of very pure solvents for sample preparation. The Uvasol<TMSYMBOL></TMSYMBOL> solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The refinement process allows a greater degree of security in applications and avoids misinterpretation of analytical results caused by traces of UV, IR and fluorescence contamination. Uvasol<TMSYMBOL></TMSYMBOL> solvents offer best UV transmittance. In all specifications the minimum transmittance for 5 typical wavelengths are identified.

Application


  • Numerical Simulation of HCCI Combustion Fuelled with Gasoline + Methyl Tert-Butyl Ether Blend.: This study uses numerical simulations to investigate the effects of adding MTBE to gasoline in homogeneous charge compression ignition (HCCI) engines, highlighting improvements in combustion efficiency and emissions reduction (Dadapeer et al., 2022).

  • INFLUENCE OF NANO CERIUM OXIDE IN EMISSION REDUCTION WITH METHYL TERT-BUTYL ETHER - GASOLINE BLENDS AT HCCI ENGINE.: This paper investigates the impact of nano cerium oxide on reducing emissions in gasoline blends with MTBE in HCCI engines, showing significant improvements in emission reduction (International Journal of Renewable Energy Research, 2023).

  • Pt/Au Nanoparticles@Co3O4 Cataluminescence Sensor for Rapid Analysis of Methyl Sec-Butyl Ether Impurity in Methyl Tert-Butyl Ether Gasoline Additive.: This research presents a sensor based on Pt/Au nanoparticles for detecting MSBE impurities in MTBE, offering a rapid and reliable method for ensuring the purity of MTBE gasoline additives (Shi et al., 2022).

Analysis Note

Purity (GC): ≥ 99.9 %
Evaporation residue: ≤ 0.0002 %
Water: ≤ 0.01 %
Colour: ≤ 10 Hazen
Acidity: ≤ 0.0002 meq/g
Alkalinity: ≤ 0.0002 meq/g
Fluorescence (as quinine at 254 nm): ≤ 1.0 ppb
Fluorescence (as quinine at 365 nm): ≤ 1.0 ppb
Transmission (at 215 nm): ≥ 40 %
Transmission (at 235 nm): ≥ 55 %
Transmission (at 240 nm): ≥ 60 %
Transmission (at 255 nm): ≥ 85 %
Transmission (at 260 nm): ≥ 90 %
Transmission (from 280 nm): ≥ 98 %
Absorbance (at 215 nm): ≤ 0.40
Absorbance (at 235 nm): ≤ 0.26
Absorbance (at 240 nm): ≤ 0.22
Absorbance (at 255 nm): ≤ 0.07
Absorbance (at 260 nm): ≤ 0.05
Absorbance (from 280 nm): ≤ 0.01
Meet the requirements of "Specific use spectroscopy" according ACS and Reag. Ph Eur

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

UVASOL is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under tert-Butyl methyl ether


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

-18.4 °F - closed cup

flash_point_c

-28 °C - closed cup

hcodes



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library