Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.01872

SAFC

Magnesium chloride hexahydrate

crystalline, Ph. Eur., JPC, USP, FCC, E 511

Pharma Manufacturing

Synonym(s):

Magnesium dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgCl2 · 6H2O
CAS Number:
Molecular Weight:
203.30
MDL number:
UNSPSC Code:
12352302
E Number:
E511
EC Index Number:
232-094-6
Pricing and availability is not currently available.

Agency

JPC
Ph. Eur.
USP

Quality Level

reg. compliance

FCC

form

crystalline
solid

pH

4.5-7.0 (20 °C, 50 g/L in H2O)

mp

117 °C (decomposition)

application(s)

liquid formulation
pharmaceutical
solid formulation

storage temp.

15-25°C

SMILES string

O.O.O.O.O.O.Cl[Mg]Cl

InChI

1S/2ClH.Mg.6H2O/h2*1H;;6*1H2/q;;+2;;;;;;/p-2

InChI key

DHRRIBDTHFBPNG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Magnesium chloride is a mineral salt which is used for a broad range of applications in pharmaceutical and food industry. In pharmaceutical formulations it is e.g. used as tonicity agent, and can serve as a strengthening agent and flavor enhancer for food and beverage.

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service