Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.01804

Supelco

Chloride Test, photometric

photometric, 0.5-15.0 mg/L (Cl-), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

Product Name

Chloride Cell Test, photometric, 0.5-15.0 mg/L (Cl-), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 25 tests

specific analyte(s)

chloride

measuring range

0.5-15.0 mg/L (Cl-)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Iron(III)-thiocyanate)

storage temp.

15-25°C

General description

Method: photometric 0.5 - 15.0 mg/l Cl⁻ Spectroquant®
This Spectroquant® Chloride test allows the accurate quantification of chloride ions in various waters and waste water. Their concentration depends on geological factors and the general local situation.
Within the test chloride ions react with mercury(II)-thiocyanate to mercury(II) chloride. The liberated thiocyanide reacts with iron ions to form a read complex. The method is analogous to EPA 325.1 and APHA 4500-Cl- E.
All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.
The Spectroquant® Cell Tests come with prefilled 16 mm round cells as well as all the required reagents to perform the analysis according to the procedure described in the accompanying instruction leaflet.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1 - STOT RE 2

Supplementary Hazards

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service