Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.01714

Supelco

Barium carbonate

for analysis EMSURE® ACS,Reag. Ph Eur

Synonym(s):

Barium carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CBaO3
CAS Number:
Molecular Weight:
197.34
MDL number:
UNSPSC Code:
12352302
EC Index Number:
208-167-3
NACRES:
NA.21
grade:
ACS reagent
form:
solid
solubility:
0.02 g/L

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

product line

EMSURE®

Assay

99.0-101.0% (BaCO3 basis, complexometric)

form

solid

potency

418 mg/kg LD50, oral (Rat)

impurities

≤0.002 meq/g Total of alkali (water soluble)
≤0.005% Oxidizing Substances (as NO3)
≤0.015% Substances insoluble in hydrochloric acid

pH

7-8 (16 °C, 0.016 g/L in H2O)

mp

>1450 °C (decomposition)

solubility

0.02 g/L

density

4.43 g/cm3 at 20 °C

bulk density

350 kg/m3

anion traces

chloride (Cl-): ≤0.002%
sulfide (S2-): ≤0.001%

cation traces

Ca: ≤0.02%
Fe: ≤0.001%
K: ≤0.005%
Na: ≤0.02%
Sr: ≤0.3%
heavy metals: ≤0.001%

storage temp.

2-30°C

SMILES string

[Ba+2].[O-]C(=O)[O-]

InChI

1S/CH2O3.Ba/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

AYJRCSIUFZENHW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Analysis Note

Assay (complexometric, BaCO₃): 99.0 - 101.0 %
Substances insoluble in hydrochloric acid: ≤ 0.015 %
Chloride (Cl): ≤ 0.002 %
Sulphide (S): ≤ 0.001 %
Heavy metals (ACS): ≤ 0.001 %
Ca (Calcium): ≤ 0.02 %
Fe (Iron): ≤ 0.001 %
K (Potassium): ≤ 0.005 %
Na (Sodium): ≤ 0.02 %
Sr (Strontium): ≤ 0.3 %
Total of alkali (water soluble): ≤ 0.002 meq/g
Oxidizing Substances (as NO₃): ≤ 0.005 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service