Skip to Content
MilliporeSigma

1.01188

Ammonium nitrate

for analysis EMSURE® ACS

Synonym(s):

Ammonium nitrate, Nitric acid ammonia, Ammonia nitrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
H4N2O3
CAS Number:
Molecular Weight:
80.04
UNSPSC Code:
12352301
EC Index Number:
229-347-8
NACRES:
NA.21
MDL number:
Assay:
≥95.0% (alkalimetric)
Grade:
ACS reagent
Form:
solid
Solubility:
1920 g/L
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

ACS reagent

Quality Segment

product line

EMSURE®

assay

≥95.0% (alkalimetric)

form

solid

potency

2462 mg/kg LD50, oral (Rat)

impurities

≤0.005% Insoluble matter

ign. residue

≤0.01% (as sulfate)

total organic carbon (TOC) residue (Combustable matter, including organic components)

≤0.2% (as carbon)

pH

4.5-7.0 (20 °C, 100 g/L in H2O)

bp

210 °C/15.0 hPa

mp

169 °C

solubility

1920 g/L

density

1.72 g/cm3 at 20 °C

bulk density

600‑700 kg/m3

anion traces

chloride (Cl-): ≤0.0003%, nitrite (NO2-): ≤0.0005%passes test, phosphate (PO43-): ≤0.0005%, sulfate (SO42-): ≤0.002%

cation traces

Ca: ≤0.003%, Fe: ≤0.0002%, % (heavy metals (ACS) ≤ 0.0005%)

storage temp.

2-30°C

SMILES string

[N+H4].[N+](=O)([O-])[O-]

InChI

1S/NO3.H3N/c2-1(3)4;/h;1H3/q-1;/p+1

InChI key

DVARTQFDIMZBAA-UHFFFAOYSA-O

Application


  • Role of gas-particle conversion of ammonia in haze pollution under ammonia-rich environment in Northern China and prospects of effective emission reduction.: This study investigates the role of ammonium nitrate in haze pollution, emphasizing its environmental impact and the potential for emission reduction strategies (Zou et al., 2024).

  • Electrocatalytic Nitrate Reduction on Metallic CoNi-Terminated Catalyst with Industrial-Level Current Density in Neutral Medium.: This paper explores the use of ammonium nitrate in electrocatalytic processes for nitrate reduction, demonstrating its application in industrial catalysis (Wei et al., 2024).

  • Thermal annealing gradient tailoring on ammonium nitrate-capped CdS nanospheres synthesized via green precipitation.: The study focuses on the synthesis of CdS nanospheres using ammonium nitrate, emphasizing its applications in nanomaterial engineering (Kok Sheng & Alrababah, 2024).

Analysis Note

Assay (alkalimetric): ≥ 95.0 %


Insoluble matter: ≤ 0.005 %


pH-value (5 %; water, 25 °C): 4.5 - 6.0


Chloride (Cl): ≤ 0.0003 %


Nitrite (NO₂):


≤ 0.0005 %


: passes test


Phosphate (PO₄): ≤ 0.0005 %


Sulfate (SO₄): ≤ 0.002 %


heavy metals (ACS) ≤ 0.0005 %


Ca (Calcium): ≤ 0.003 %


Fe (Iron): ≤ 0.0002 %


Residue on ignition (as sulfate): ≤ 0.01 %


The following parameter from a double TOC determination of one batch:


Combustible matter, including organic components (as carbon) ≤ 0.2 %


(meets the specified value)


In accordance with class 5.1 UN 1942.

Other Notes

Replaces: AX1315-1

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Ammonium nitrate


pictograms

Flame over circleExclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 3

Storage Class

5.1C - Ammonium nitrate and ammonium nitrate containing preparations

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library