Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.01057

Supelco

Aluminium

(foil) for analysis 0.3 mm thickness, 30 mm width EMSURE®

Synonym(s):

Aluminium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Al
CAS Number:
Molecular Weight:
26.98
MDL number:
UNSPSC Code:
12141702
EC Index Number:
231-072-3
NACRES:
NA.21

Quality Level

product line

EMSURE®

form

solid

impurities

≤0.0002% As (Arsenic)
≤0.002% Mn (Manganese)
≤0.005% Cu (Copper)
≤0.005% Total nitrogen (N)
≤0.005% Zn (Zinc)
≤0.05% Fe (Iron)
≤0.06% Si (Silicon)

bp

2467 °C/1013 hPa

mp

660 °C

density

2.70 g/cm3 at 20 °C

storage temp.

no temp limit

SMILES string

[Al]

InChI

1S/Al

InChI key

XAGFODPZIPBFFR-UHFFFAOYSA-N

Related Categories

Application


  • Sensitive substrates for surface-enhanced Raman scattering: Presents substrates based on gold and silver nanoparticles, with aluminum used to improve the substrates′ resistance to laser damage. This development supports researchers in chemistry and material sciences focusing on analytical techniques (Mayr F et al., 2023).

Analysis Note

Total nitrogen (N): ≤ 0.005 %
As (Arsenic): ≤ 0.0002 %
Cu (Copper): ≤ 0.005 %
Fe (Iron): ≤ 0.05 %
Mn (Manganese): ≤ 0.002 %
Si (Silicon): ≤ 0.06 %
Zn (Zinc): ≤ 0.005 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service