Skip to Content

1.00987

Benzyl alcohol

EMPROVE® EXPERT, Ph. Eur., BP, ChP, JP, NF

Synonym(s):

Benzenemethanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H5CH2OH
CAS Number:
Molecular Weight:
108.14
EC Index Number:
202-859-9
UNSPSC Code:
12352104
Beilstein/REAXYS Number:
878307
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


agency

ChP, JP, NF, Ph. Eur., BP

Quality Segment

vapor density

3.7 (vs air)

vapor pressure

0.07 hPa ( 20 °C), 13.3 mmHg ( 100 °C), 3.75 mmHg ( 77 °C)

product line

EMPROVE® EXPERT

assay

98.0-100.5% (GC)

form

liquid

autoignition temp.

435 °C (DIN 51794), 817 °F

refractive index

n20/D 1.539 (lit.)

bp

203-205 °C (lit.)

mp

−16-−13 °C (lit.)

transition temp

flash point 101 °C (DIN 51758)

density

1.045 g/mL at 25 °C (lit.)

application(s)

liquid formulation
ophthalmics
parenterals
pharma/biopharma processes
pharmaceutical

storage temp.

2-25°C

SMILES string

OCc1ccccc1

InChI

1S/C7H8O/c8-6-7-4-2-1-3-5-7/h1-5,8H,6H2

InChI key

WVDDGKGOMKODPV-UHFFFAOYSA-N

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Benzyl alcohol is typically used as a preservative in pharmaceutical applications and is active against bacteria, yeast and mould. It can also serve as a solvent for poorly soluble drugs.

Benzyl Alcohol Emprove® Expert is an extraordinarily pure product with impurity specification thresholds even below current requirements of designated pharmacopoeias. Therefore, this product is especially suitable for high-risk applications or any other purpose that requires superior quality.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Benzyl alcohol


pictograms

Exclamation mark

signalword

Warning

Storage Class

10 - Combustible liquids

flash_point_f

213.8 °F - DIN 51758

flash_point_c

101 °C - DIN 51758

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1B



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.