Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

1.00921

Supelco

Diethyl ether

≥99.7% (GC), ACS reagent, contains 5-10 ppm 2, 6-Di-tert-butyl-4-methylphenol (BHT) as stabilizer, reag. Ph. Eur., reag. ISO, EMSURE®

Synonym(s):

Et2O, Ethoxyethane, Ethyl ether, Ether, Ether, Ethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2)2O
CAS Number:
Molecular Weight:
74.12
Beilstein:
1696894
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-467-2
NACRES:
NA.05
grade:
ACS reagent
for analytical purposes
bp:
34.6 °C (lit.)
vapor pressure:
587 hPa ( 20 °C)

Product Name

Diethyl ether, for analysis EMSURE® ACS,ISO,Reag. Ph Eur

grade

ACS reagent
for analytical purposes

Quality Level

Agency

reag. ISO
reag. Ph. Eur.

vapor density

2.6 (vs air)

vapor pressure

587 hPa ( 20 °C)

product line

EMSURE®

Assay

≥99.7% (GC)

form

liquid

autoignition temp.

320 °F

potency

1215 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

contains

5-10 ppm 2,6-Di-tert-butyl-4-methylphenol (BHT) as stabilizer

expl. lim.

36.5 %

dilution

(for analytical testing)

impurities

≤0.000015% Peroxide
≤0.00003% Chloride (Cl)
≤0.00003% Sulfate (SO4)
≤0.0002 meq/g Alkalinity
≤0.0002 meq/g Titrable acid
≤0.001% Carbonyl compounds (as CO)
≤0.001% Carbonyl compounds (as HCHO)
≤0.005% Acetone (GC)
≤0.02% Ethanol (GC)
≤0.02% Methanol (GC)
≤0.03% Water

evapn. residue

≤0.0005%

color

clearAPHA: ≤10

refractive index

n20/D 1.3530 (lit.)

bp

34.6 °C (lit.)

mp

−116 °C (lit.)

transition temp

flash point -40 °C

density

0.706 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.00005%
B: ≤0.000002%
Ba: ≤0.00001%
Ca: ≤0.00005%
Cd: ≤0.000005%
Co: ≤0.000002%
Cr: ≤0.000002%
Cu: ≤0.000002%
Fe: ≤0.00001%
Mg: ≤0.00001%
Mn: ≤0.000002%
Ni: ≤0.000002%
Pb: ≤0.00001%
Sn: ≤0.00001%
Zn: ≤0.00001%

SMILES string

CCOCC

storage temp.

15-25°C

InChI

1S/C4H10O/c1-3-5-4-2/h3-4H2,1-2H3

InChI key

RTZKZFJDLAIYFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

EMSURE grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE provides worldwide best and most extensive product specifications. We declare our EMSURE range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

Application

Diethyl ether is used as a solvent:
  • For the solid-phase extraction (SPE) process.
  • To wash strata Si-1 cartridges.

Linkage

Discover EMSURE - to be more than sure!

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 1 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-40.0 °F - closed cup

Flash Point(C)

-40 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of coenzyme Q10, alpha-tocopherol and ascorbic acid on oxidation of cholesterol in chicken liver pate
T Polak, et al.
LWT--Food Science and Technology, 44, 1052-1058 (2011)

Articles

EMSURE® Grade Inorganic Solvents and Reagents offer exceptional purity and quality control, ensuring reliable, compliant results for regulated analyses.

EMSURE® Grade Inorganic Solvents and Reagents offer exceptional purity and quality control, ensuring reliable, compliant results for regulated analyses.

EMSURE® Grade Inorganic Solvents and Reagents offer exceptional purity and quality control, ensuring reliable, compliant results for regulated analyses.

EMSURE® Grade Inorganic Solvents and Reagents offer exceptional purity and quality control, ensuring reliable, compliant results for regulated analyses.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service