Skip to Content

1.00894

Sucralose

powder, EMPROVE® EXPERT, Ph. Eur., ChP, JPE, NF

Synonym(s):

Sucralose, 1,6-Dichloro-1,6-dideoxy-β-D-fructofuranosyl-4-chloro-4-deoxy-α-D-galactopyranoside, E955, Trichlorosucrose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C12H19Cl3O8
CAS Number:
Molecular Weight:
397.63
EC Index Number:
259-952-2
UNSPSC Code:
12352121
Beilstein/REAXYS Number:
3654410
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

ChP, JPE, NF, Ph. Eur.

description

EMPROVE® EXPERT Ph Eur,NF,JPE

product line

EMPROVE® EXPERT

form

fine powder, powder

reaction suitability

reaction type: Olefin Metathesis

particle size

≤12 μm (d90)

pH

6-8 (20 °C, 100 g/L in H2O)

mp

130 °C (decomposition)

density

1.62 g/cm3 at 20 °C

application(s)

continuous manufacturing
liquid formulation
pharmaceutical
solid formulation

storage temp.

15-25°C

SMILES string

OC[C@H]1O[C@H](O[C@]2(CCl)O[C@H](CCl)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@H]1Cl

InChI

1S/C12H19Cl3O8/c13-1-4-7(17)10(20)12(3-14,22-4)23-11-9(19)8(18)6(15)5(2-16)21-11/h4-11,16-20H,1-3H2/t4-,5-,6+,7-,8+,9-,10+,11-,12+/m1/s1

InChI key

BAQAVOSOZGMPRM-QBMZZYIRSA-N

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our EMPROVE® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Sucralose is a high-intensity sweetener which is non-cariogenic and suitable for diabetics and patients with an impaired glucose tolerance. In addition, it is readily soluble and masks both bitterness and medicinal off-notes with equal ease – making it exceptionally well-suited for pharmaceutical applications.
Our Sucralose is available in micronized or granular form, both pharmaceutical grade, allowing you to choose depending on your individual needs. Due to its optimal dispersion behavior, sucralose powder is typically recommended for solid dose formulations, while the fast dissolution behavior of sucralose granular makes it very well-suited for liquid formulations.

Biochem/physiol Actions

A synthetic sweet tastant detectable by humans. Activates T1R2/T1R3 sweet taste receptors on enteroendocrine cells and elicits increased hormonal secretion of glucagon-like peptide-1 and glucose-dependent insulinotrophic peptide.

Features and Benefits

  • Excellent sugar-like taste profiles
  • Masking bitter aftertaste and medical off-notes with ease
  • High sweetness potency offers improved cost-efficiency
  • Enhanced stability
  • Non-cariogenic and suitable for diabetics and patients with phenylketonuria

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

The substance 1.00894 has no REACH registration because the only supported use is the use in medicinal products for human or veterinary use or in food or feedingstuffs according to article 2 of the REACH Regulation (EC) No 1907/2006.


Still not finding the right product?

Explore all of our products under Sucralose


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.