Skip to Content
MilliporeSigma

Skip To

1.00808

Titanium dioxide

for analysis EMSURE® Reag. Ph Eur

Synonym(s):

Titanium(IV) oxide, Titanic anhydride, Titanium dioxide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
TiO2
CAS Number:
Molecular Weight:
79.87
UNSPSC Code:
12352303
EC Index Number:
236-675-5
NACRES:
NA.21
MDL number:
Assay:
≥99.0% dry basis (cerimetric), 98.0-100.5% (cerimetric)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Titanium(IV) oxide, for analysis EMSURE® Reag. Ph Eur

agency

reag. Ph. Eur.

Quality Segment

product line

EMSURE®

assay

≥99.0% dry basis (cerimetric), 98.0-100.5% (cerimetric)

form

powder

potency

>10000 mg/kg LD50, oral (Rat)

impurities

≤0.5% Substances soluble in dilute hydrochloric acid, ≤0.5% Substances soluble in water

loss

≤ 0.5% loss on drying, 105 °C; 3 h

pH

7-8 (20 °C, 100 g/L in H2O, slurry)

bp

2900 °C/1013 hPa

mp

1855 °C

bulk density

850 kg/m3

cation traces

As: ≤0.0001%, Ba: ≤0.0020%, Fe: ≤0.005%, Pb: ≤0.0005%, Sb: ≤0.0020%, Zn: ≤0.005%

storage temp.

2-30°C

SMILES string

[Ti](=O)=O

InChI

1S/2O.Ti

InChI key

GWEVSGVZZGPLCZ-UHFFFAOYSA-N

Application


  • Sulfanyl Porphyrazines with Morpholinylethyl Periphery-Synthesis, Electrochemistry, and Photocatalytic Studies after Deposition on Titanium(IV) Oxide P25 Nanoparticles.: This study presents the synthesis and characterization of sulfanyl porphyrazines with morpholinylethyl peripherals. These compounds were deposited on Titanium(IV) oxide nanoparticles and evaluated for their photocatalytic properties, indicating potential applications in environmental photocatalysis and organic synthesis (Koczorowski et al., 2021).

  • Gold-titanium(IV) oxide plasmonic photocatalysts prepared by a colloid-photodeposition method: correlation between physical properties and photocatalytic activities.: This research discusses the development of gold-Titanium(IV) oxide plasmonic photocatalysts using a colloid-photodeposition method. It explores the correlation between their physical properties and enhanced photocatalytic activities, suggesting applications in solar energy conversion and environmental remediation (Tanaka et al., 2012).

  • Size-selective photocatalytic reactions by titanium(IV) oxide coated with a hollow silica shell in aqueous solutions.: This paper explores the use of Titanium(IV) oxide coated with hollow silica shells for size-selective photocatalytic reactions. The unique structural properties of these composites allow for selective photocatalytic degradation of organic pollutants in water, providing a novel approach to water purification (Ikeda et al., 2007).

  • Photocatalytic activity of neat and silicon-doped titanium(IV) oxide nanofibers prepared by combined sol-gel and electrospinning techniques.: Investigates the photocatalytic performance of silicon-doped Titanium(IV) oxide nanofibers. These nanofibers show enhanced photocatalytic activity due to increased surface area and modified electronic properties, making them effective for pollution control and solar energy applications (Watthanaarun et al., 2007).

  • Enhanced photocatalysis of pentachlorophenol by metal-modified titanium (IV) oxide.: Focuses on the photocatalytic degradation of pentachlorophenol using metal-modified Titanium(IV) oxide. The study demonstrates that the modification with metals significantly enhances the photocatalytic efficiency, offering a potent method for the detoxification of hazardous organic compounds in environmental applications (Lin et al., 2006).

Analysis Note

Assay (cerimetric): 98.0 - 100.5 %
Assay (cerimetric, calculated on dried substance): ≥ 99.0 %
Identity: passes test
Appearance of solution: passes test
Substances soluble in water: ≤ 0.5 %
Substances soluble in dilute hydrochloric acid: ≤ 0.5 %
Acidity or alkalinity: passes test
As (Arsenic): ≤ 0.0001 %
Ba (Barium): ≤ 0.0020 %
Fe (Iron): ≤ 0.005 %
Pb (Lead): ≤ 0.0005 %
Sb (Antimony): ≤ 0.0020 %
Zn (Zinc): ≤ 0.005 %
Loss on drying (105 °C; 3h): ≤ 0.5 %

Other Notes

Replaces: TX0685; TX0685-2

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
10080514021193803
description

for analysis EMSURE® Reag. Ph Eur

description

EMPROVE® ESSENTIAL, Ph. Eur., BP, ChP, JP, USP, E 171

description

ReagentPlus®, ≥99%

description

EMPLURA®

form

powder

form

fine powder

form

nanoparticles

form

-

assay

≥99.0% dry basis (cerimetric), 98.0-100.5% (cerimetric)

assay

-

assay

≥99%

assay

-

Quality Level

300

Quality Level

500

Quality Level

-

Quality Level

-

pH

7-8 (20 °C, 100 g/L in H2O, slurry)

pH

7-8 (20 °C, 100 g/L in H2O, slurry)

pH

-

pH

-

cation traces

As: ≤0.0001%, Fe: ≤0.005%, Sb: ≤0.0020%, Ba: ≤0.0020%, Zn: ≤0.005%, Pb: ≤0.0005%

cation traces

-

cation traces

As: ≤5 mg/kg, Fe: ≤50 mg/kg, Pb: ≤50 mg/kg

cation traces

-

mp

1855 °C

mp

1855 °C

mp

>350 °C (lit.)

mp

-


Still not finding the right product?

Explore all of our products under Titanium(IV) oxide


Storage Class

13 - Non Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. Is this P25?

    1 answer
    1. This product is not characterized by form (anatase, rutile, or a mixture); therefore, it cannot be classified as Aeroxide P25. TiO2 Aeroxide P25 is composed of anatase and rutile crystallites, the reported ratio being typically 70:30 or 80:20.

      Helpful?

  2. Hello, I want to know what is the size of TiO2 particles in the product with CAS 13463-67-7. Thank you!

    1 answer
    1. The particle size of this product has not been determined on a lot to lot basis. However, historical data suggests a particle size distribution as follows:
      d(10): 0.4 µm (up to 10%)
      d(50): 0.7 µm (up to 50%)
      d(90): 1.3 µm (up to 90%)

      Helpful?

Reviews

No rating value

Active Filters