Skip to Content
MilliporeSigma

1.00807

Trichloroacetic acid

for analysis EMSURE® ACS,Reag. Ph Eur

Synonym(s):

Trichloroacetic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C2HCl3O2
CAS Number:
Molecular Weight:
163.39
UNSPSC Code:
12352106
EC Index Number:
200-927-2
NACRES:
NA.21
MDL number:
Assay:
≥99.5% (alkalimetric)
Concentration:
≥99.5%
Form:
solid
Grade:
ACS reagent
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

ACS reagent

Quality Segment

agency

reag. Ph. Eur.

vapor pressure

1 hPa ( 20 °C)

product line

EMSURE®

assay

≥99.5% (alkalimetric)

form

solid

autoignition temp.

711 °C

potency

3320 mg/kg LD50, oral (Rat)

concentration

≥99.5%

impurities

≤0.01% In water insoluble matter, ≤300 ppm Sulfated ash (600 °C)

color

clear

pH

<1 (20 °C, 50 g/L in H2O)

mp

54-56 °C

transition temp

flash point >110 °C

solubility

1300 g/L

density

1.63 g/cm3 at 20 °C

bulk density

900 kg/m3

anion traces

chloride (Cl-): ≤10 ppm, nitrate (NO3-): ≤20 ppm, phosphate (PO43-): ≤5 ppm, sulfate (SO42-): ≤200 ppm

cation traces

Cu: ≤5 ppm, Fe: ≤10 ppm, heavy metals (as Pb): ≤20 ppm

storage temp.

15-25°C

SMILES string

ClC(Cl)(Cl)C(=O)[O-].[H+]

InChI

1S/C2HCl3O2/c3-2(4,5)1(6)7/h(H,6,7)

InChI key

YNJBWRMUSHSURL-UHFFFAOYSA-N

Application

Trichloroacetic acid (TCA) is used in:
  • Ascorbic acid assay to homogenize microalgae samples under ice-cold conditions, facilitating the extraction of ascorbic acid (vitamin C).
  • Volatile Basic Nitrogen (VBN) assay to extract volatile basic nitrogen from the sample, aiding in the assessment of the freshness and quality of the meat.

Analysis Note

Assay (alkalimetric): ≥ 99.5 %
Identity (IR-spectrum): passes test
Appearance: colorless, deliquescent crystals
Appearance of solution (200 g/l; water): clear and colorless
In water insoluble matter: ≤ 0.01 %
Chloride (Cl): ≤ 10 ppm
Nitrate (NO₃): ≤ 20 ppm
Phosphate (PO₄): ≤ 5 ppm
Sulphate (SO₄): ≤ 200 ppm
Heavy metals (as Pb): ≤ 20 ppm
Cu (Copper): ≤ 5 ppm
Fe (Iron): ≤ 10 ppm
Readily carbonisable substance: passes test
Sulfated ash (600 °C): ≤ 300 ppm

Other Notes

Replaces: TX1045; TX1045-5; TX1045-3

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Trichloroacetic acid


pictograms

CorrosionEnvironment

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1A

Storage Class

8B - Non-combustible, corrosive hazardous materials

wgk

WGK 2

flash_point_f

235.4 °F - closed cup

flash_point_c

> 113 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library