Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.00700

Supelco

Thioglycolic acid

GR for analysis Reag. Ph Eur

Synonym(s):

Thioglycolic acid, Mercaptoacetic acid, 2-Mercaptoethanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H4O2S
CAS Number:
Molecular Weight:
92.12
MDL number:
UNSPSC Code:
12352106
EC Index Number:
200-677-4
NACRES:
NA.21

Agency

reag. Ph. Eur.

Quality Level

vapor pressure

0.1 hPa ( 20 °C)

potency

73 mg/kg LD50, oral (Rat)

pH

1 (20 °C in H2O)

bp

220 °C/1013 hPa (decomposition)

mp

-16.5 °C

transition temp

flash point 131.5 °C

density

1.325 g/cm3 at 20 °C

storage temp.

2-8°C

InChI

1S/C2H4O2S/c3-2(4)1-5/h5H,1H2,(H,3,4)

InChI key

CWERGRDVMFNCDR-UHFFFAOYSA-N

Application


  • Low-Temperature Synthesis of Bi(2)S(3) Hierarchical Microstructures via Co-Precipitation and Digestive Process in Aqueous Medium: This study highlights the use of thioglycolic acid in the synthesis of bismuth sulfide microstructures, demonstrating its role in facilitating low-temperature reactions, which is crucial for the development of advanced materials in various industrial applications (Carrasco-González et al., 2024).

  • CdS Semiconductor Quantum Dots; Facile Synthesis, Application as Off Fluorescent Sensor for Detection of Lead (Pb(2+)) Ions and Catalyst for Degradation of Dyes from Water: The study explores thioglycolic acid′s role in synthesizing quantum dots for environmental monitoring and purification systems, reflecting its critical application in environmental safety and nanotechnology (Farahmandzadeh et al., 2023).

Analysis Note

Description: clear, colourless liquid
Identity: conforms
Assay (iodometric): ≥ 99 %
Solubility (USP): conforms
Sensitivity (USP): conforms
Fe (Iron): ≤ 0.00005 %
Organic bound chlorine: ≤ 0.01 %
Sulfated ash: ≤ 0.05 %
The statement of the assay applies to the test date.Agradual substance-specific content reduction,especially at higher temperatures,must be expected.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

266.0 °F - closed cup

Flash Point(C)

130 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service