Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.00583

Supelco

Tungstophosphoric acid hydrate

for analysis EMSURE®

Synonym(s):

Tungstophosphoric acid hydrate, Phosphotungstic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3[P(W3O10)4] · x H2O
CAS Number:
Molecular Weight:
2880.05 (anhydrous basis)
MDL number:
UNSPSC Code:
12352301
EC Index Number:
215-682-7
NACRES:
NA.21

Quality Level

product line

EMSURE®

form

solid

potency

3300 mg/kg LD50, oral (Rat)

loss

≤17% loss on ignition, 750 °C

pH

1.3 (20 °C, 20 g/L in H2O)

mp

107 °C

solubility

water: soluble at 20 °C

bulk density

960 kg/m3

anion traces

chloride (Cl-): ≤50 ppm
sulfate (SO42-): ≤100 ppm

cation traces

Cu: ≤10 ppm
Fe: ≤20 ppm
K: ≤200 ppm
Na: ≤200 ppm
Pb: ≤20 ppm

storage temp.

15-25°C

SMILES string

[W](=O)(=O)=O.[W](=O)(=O)=O.[W](=O)(=O)=O.[W](=O)(=O)=O.[W](=O)(=O)=O.[W](=O)(=O)=O.[W](=O)(=O)=O.[W](=O)(=O)=O.[W](=O)(=O)=O.[W](=O)(=O)=O.[W](=O)(=O)=O.[W](=O)(=O)=O.[P](=O)(O)(O)O.O

InChI

1S/H3O4P.H2O.36O.12W/c1-5(2,3)4;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/h(H3,1,2,3,4);1H2;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;

InChI key

AVFBYUADVDVJQL-UHFFFAOYSA-N

Application


  • Calixarene based portable sensor for the direct assay of indiscriminate ephedrine content of weight loss herbal preparations.: Tungstophosphoric acid hydrate is utilized as a proton conductor in the development of a calixarene-based sensor, enabling precise detection of ephedrine in herbal products, showcasing its application in analytical chemistry for public health safety (Wahba et al., 2021).

  • A New Solid-State Proton Conductor: The Salt Hydrate Based on Imidazolium and 12-Tungstophosphate.: This article presents tungstophosphoric acid hydrate as a key component in a novel solid-state proton conductor, offering significant advancements in materials science for fuel cells and other energy applications (Martinelli et al., 2021).

  • Location, acid strength, and mobility of the acidic protons in Keggin 12-H3PW12O40: a combined solid-state NMR spectroscopy and DFT quantum chemical calculation study.: The research investigates the acidic properties and proton mobility of tungstophosphoric acid hydrate using advanced NMR and DFT techniques, providing valuable insights for catalysis and materials science (Yang et al., 2005).

Linkage

Replaces: PX1055; PX1055-4

Analysis Note

Chloride (Cl): ≤ 50 ppm
Sulfate (SO₄): ≤ 100 ppm
Cu (Copper): ≤ 10 ppm
Fe (Iron): ≤ 20 ppm
K (Potassium): ≤ 200 ppm
Na (Sodium): ≤ 200 ppm
Pb (Lead): ≤ 20 ppm
Loss on ignition (750 °C): ≤ 17 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service