Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.00358

Supelco

di-Iodine pentoxide

for analysis granular 0.5-2.5 mm EMSURE®

Synonym(s):

di-Iodine pentoxide, Diiodine pentoxide, Iodine(V) oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
I2O5
CAS Number:
Molecular Weight:
333.81
MDL number:
UNSPSC Code:
12352303
EC Index Number:
234-740-2
NACRES:
NA.21

Quality Level

product line

EMSURE®

pH

1.1 (25 °C, 100 g/L in H2O)

density

4.98 g/cm3 at 20 °C

bulk density

2000 kg/m3

storage temp.

2-30°C

SMILES string

[I](=O)(=O)O[I](=O)=O

InChI

1S/I2O5/c3-1(4)7-2(5)6

InChI key

BIZCJSDBWZTASZ-UHFFFAOYSA-N

Analysis Note

Assay (iodometric, I₂O₅): ≥ 96 %
Heavy metals (as Pb): ≤ 0.001 %
Fe (Iron): ≤ 0.001 %
Loss on drying (200 °C): ≤ 2 %
Residue on ignition (as sulfates): ≤ 0.02 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Ox. Sol. 2 - Skin Corr. 1B

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service