Skip to Content
MilliporeSigma

Key Documents

Skip To

1.00286

L-Glutamine

EMPROVE® EXPERT, DAB, JP, USP

Synonym(s):

L-Glutamic acid-5-amide, Gln, (S)-2,5-Diamino-5-oxopentanoic acid, L-Glutamic acid 5-amide, Levoglutamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
H2NCOCH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
146.14
UNSPSC Code:
12352209
EC Index Number:
200-292-1
Beilstein/REAXYS Number:
1723797
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

DAB, JP, USP

description

EMPROVE® EXPERT DAB,USP

product line

EMPROVE® EXPERT

form

solid

potency

7500 mg/kg LD50, oral (Rat)

technique(s)

API processing | cocrystal formation: suitable, API processing | salt formation: suitable

pH

4.0-6.0 (20 °C, 25 g/L in H2O)

mp

185 °C (dec.) (lit.)

solubility

100 g/L

density

1.47 g/cm3 at 20 °C

bulk density

640 kg/m3

application(s)

liquid formulation

storage temp.

2-30°C

SMILES string

N[C@@H](CCC(N)=O)C(O)=O

InChI

1S/C5H10N2O3/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H2,7,8)(H,9,10)/t3-/m0/s1

InChI key

ZDXPYRJPNDTMRX-VKHMYHEASA-N

General description

Our SAFC® portfolio of high-quality products for biopharmaceutical processing withstands strict quality control procedures and is produced according to MQ-500 requirements as defined by the M-Clarity program.

M-Clarity Program

As part of our EMPROVE® Program, our raw materials are offered with EMPROVE® Dossiers which provide comprehensive, up-to-date documentation to help you navigate regulatory challenges, manage risks, and improve your manufacturing processes.

Our comprehensive portfolio of upstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Trust us to deliver supply chain transparency and reliable sourcing around the globe, streamlining your product qualification with best-in-class regulatory support and service.

Biochem/physiol Actions

L-Glutamine is an essential amino acid that is a crucial component of culture media that serves as a major energy source for cells in culture. L-Glutamine is very stable as a dry powder and as a frozen solution. In liquid media or stock solutions, however, L-glutamine degrades relatively rapidly. Optimal cell performance usually requires supplementation of the media with L-glutamine prior to use.

Analysis Note

Assay (perchloric acid titration, calculated on dried substance)99.0 - 101.0 %Identity (IR-spectrum)passes testAppearancewhite to almost white, fine powderAppearance of solution (25 g/l; CO₂-free water)clear and colorlessSpec. rotation (α 20/D, 100 g/l, hydrochloric acid 2 mol/l, calc. on dried substance)+31.5 to +33.0Spec. rotation (α 20/D, 40 g/l, water, calc. on dried substance)+6.3 to +7.3pH (25 g/l; CO₂-free water)4.0 - 6.0Chloride (Cl)≤ 200 ppmSulfate (SO₄)≤ 200 ppmHeavy metals (as Pb)≤ 10 ppmCu (Copper)≤ 25 ppmFe (Iron)≤ 10 ppmFe (Iron) (ICP)≤ 10 ppmMn (Manganese)≤ 25 ppmNi (Nickel)≤ 2 ppmZn (Zinc)≤ 130 ppmNH₄ (Ammonium)≤ 0.1 %Ninhydrin - positive substances (TLC)≤ 0.5 %Residual solvents (ICH Q3C)excluded by manufacturing processSulfated ash (600 °C)≤ 0.1 %Loss on drying (105 °C; 3 h)≤ 0.3 %Bacterial endotoxins≤ 2.0 I.U./gElemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).Corresponds to DAB, USP

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under L-Glutamine


wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

11 - Combustible Solids



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.



Questions

Reviews

No rating value

Active Filters