Skip to Content
MilliporeSigma

Skip To

1.00238

Sodium hydroxide solution

50%, EMPROVE® EXPERT

Synonym(s):

Natriumhydroxidlösung

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352305
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Sodium hydroxide solution 50%, EMPROVE® EXPERT

vapor pressure

2 hPa ( 20 °C)

Quality Segment

product line

EMPROVE® EXPERT

form

liquid

pH

14 (20 °C, 89 g/L in H2O)

bp

135 °C/1013 hPa

mp

12 °C

density

1.52 g/cm3 at 20 °C

storage temp.

15-25°C

General description

EMPROVE® EXPERT

Analysis Note

Assay (acidimetric,NaOH)49.0 - 50.0 %Carbonate (as Na₂CO₃)≤ 1.0 %Chloride (Cl)≤ 100 ppmSulfate (SO₄)≤ 30 ppmHeavy metals (as Pb)≤ 5.0 ppmAl (Aluminium)≤ 10 ppmFe (Iron)≤ 5 ppmNi (Nickel) *≤ 2.00 ppmResidual solvents (ICH Q3C)...excluded by production processFiltered with 0.2 µm
This product has been manufactured from Sodium hydroxide pellets Ph Eur, BP, JP, NF and water for injection (wfi) acc. USP which conforms with highly purified water (hpw) acc. Ph Eur.
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.055891.587931.00232
form

liquid

form

liquid

form

liquid

form

liquid

product line

EMPROVE® EXPERT

product line

-

product line

EMSURE®

product line

EMPROVE® EXPERT

vapor pressure

2 hPa ( 20 °C)

vapor pressure

3 mmHg ( 37 °C)

vapor pressure

2 hPa ( 20 °C), 3 mmHg ( 37 °C)

vapor pressure

-

pH

14 (20 °C, 89 g/L in H2O)

pH

14 (20 °C in H2O)

pH

14 (20 °C, 89 g/L in H2O)

pH

13.8 (20 °C in H2O)

Quality Level

500

Quality Level

100

Quality Level

300

Quality Level

500

bp

135 °C/1013 hPa

bp

-

bp

135 °C/1013 hPa

bp

-


Still not finding the right product?

Explore all of our products under Sodium hydroxide solution 50%


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters