Skip to Content
MilliporeSigma

1.00212

Selenite Cystine Broth

GranuCult® prime, according to ISO 19250, FDA-BAM, AOAC®, APHA, granular, selective for Salmonella spp., pack of 500 g

Synonym(s):

Salmonella selective broth, Selenite Cystine Medium, Selenite-cystine broth

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.74
UNSPSC Code:
41106213
Manufacturer/tradename:
GranuCult® prime
Agency:
AOAC®, APHA, FDA-BAM, according to ISO 19250
Ph:
6.8-7.2 (37 °C, 23 g/L in H2O)
Form:
granular
Suitability:
selective for Salmonella spp.
Application(s):
clinical
food and beverages
industrial qc
microbiology
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

AOAC®, APHA, FDA-BAM, according to ISO 19250

sterility

non-sterile

form

granular

packaging

pack of 500 g

manufacturer/tradename

GranuCult® prime

technique(s)

microbiological culture: suitable

pH

6.8-7.2 (37 °C, 23 g/L in H2O)

solubility

23 g/L

bulk density

900 kg/m3

application(s)

clinical
food and beverages
industrial qc
microbiology

storage temp.

2-8°C

suitability

selective for Salmonella spp.

General description

GranuCult® prime Selenite Cystine Broth is for the selective enrichment of Salmonella from food and animal feed and other materials, especially when the detection of Salmonella enterica subspecies enterica serovars Typhi and Paratyphi is of specific concern.
Peptones including the enzymatic digest of casein provide nitrogen, vitamins, amino acids and Lactose provides the carbon sources. Disodium phosphate acts as a buffer to maintain the pH.
Selenite inhibits the growth of coliform bacteria and enterococci in the first 6-12 hours of incubation, its inhibitory effect then gradually declines.
L-Cystine is considered to enhance the growth of Salmonella by reduction of toxicity. According to the modification described by North and Bartram (1953), L-Cystine is added to the original formulation of Leifson (1936).

Application

GranuCult® prime Selenite Cystine Broth is used for the selective enrichment of Salmonella from food and animal feed and other materials, especially when the detection of Salmonella enterica subspecies enterica serovars Typhi and Paratyphi is of specific concern.

Other Notes

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

AOAC is a registered trademark of AOAC International
GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Selenite Cystine Broth


pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

supp_hazards

Storage Class

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library