Skip to Content
MilliporeSigma

1.00191

Bismuth Sulfite (BS) Agar

acc. to Wilson and Blair, NutriSelect® Prime, powder, pack of 500 g, suitable for microbiology

Synonym(s):

BSA, WILSON BLAIR Agar, Wismuth sulfite agar

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.74
UNSPSC Code:
41106213
Manufacturer/tradename:
NutriSelect® Prime
Agency:
ISO 6579-1, USP 31
Ph:
7.7±0.2 (25 °C)
Form:
powder
Suitability:
selective and differential for enterobacteriaceae: Salmonella spp.
Application(s):
food and beverages
pharmaceutical, microbiology
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

ISO 6579-1, USP 31

description

FDA-BAM

form

powder

packaging

pack of 500 g

manufacturer/tradename

NutriSelect® Prime

technique(s)

microbiological culture: suitable

pH

7.7±0.2 (25 °C)

application(s)

food and beverages
pharmaceutical, microbiology

storage temp.

2-8°C

suitability

selective and differential for enterobacteriaceae: Salmonella spp.

General description

NutriSelect® Bismuth Sulfite (BS) Agar introduced by Wilson-Blair (1927) is specifically designed for the isolation and differentiation of Salmonella spp from food, animal feed and other materials precisely when the detection of Salmonella enterica subspecies enterica serovars Typhi and Paratyphi is of concern.
Colonies of H2S positive Salmonella exhibit blackening due to the formation of iron sulfide. The media selectivity is enhanced by the presence of brilliant green and bismuth sulfite which inhibit the growth of gram-positive bacteria and coliforms in the sample.
Peptone, meat extract and glucose allow good growth. Disodium phosphate acts as a buffer to maintain pH and agar is the solidifying agent.

Application

NutriSelect® Bismuth Sulfite (BS) Agar is used to isolate and differentiate Salmonella spp. from food animal feed and other materials. The medium is used to selectively grow the species when S. typhi and S. paratyphi are expected.

Analysis Note

Salmonella colonies display blackening after 18 hrs of incubation.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs. The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.


hcodes

Hazard Classifications

Aquatic Chronic 3

Storage Class

11 - Combustible Solids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library