Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.00132

Supelco

Barbituric acid

for analysis EMSURE®

Synonym(s):

Barbituric acid, 2,4,6-Trihydroxypyrimidine, N,N′-Malonylurea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4N2O3
CAS Number:
Molecular Weight:
128.09
MDL number:
UNSPSC Code:
12352115
EC Index Number:
200-658-0
NACRES:
NA.21

Quality Level

product line

EMSURE®

Assay

≥99% (acidimetric)

form

solid

potency

>5000 mg/kg LD50, oral (Rat)

sulfated ash

≤0.1%

loss

≤0.1% loss on drying, 105°C

pH

2-3 (60 °C, 50 g/L in H2O)

mp

250-252 °C (decomposition)

solubility

11.45 g/L

bulk density

720 kg/m3

anion traces

chloride (Cl-): ≤40 ppm

cation traces

Fe: ≤10 ppm
heavy metals (as Pb): ≤50 ppm

storage temp.

15-25°C

InChI

1S/C4H4N2O3/c7-2-1-3(8)6-4(9)5-2/h1H2,(H2,5,6,7,8,9)

InChI key

HNYOPLTXPVRDBG-UHFFFAOYSA-N

Linkage

Replaces: BX0035; BX0035-3

Analysis Note

Assay (acidimetric): ≥ 99
Identity (IR-spectrum): passes test
Chloride (Cl): ≤ 40 ppm
Heavy metals (as Pb): ≤ 50 ppm
Fe (Iron): ≤ 10 ppm
Sulfated ash: ≤ 0.1 %
Loss on Drying (105°C): ≤ 0.1 %
Suitability as reagent (for cyanide determination): passes test

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150.00 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service