Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.00126

Millipore

L-Aspartic acid

for biochemistry

Synonym(s):

L-Aspartic acid, L-α-Aminosuccinic acid, L-αlpha-Aminosuccinic acid, Asp

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7NO4
CAS Number:
Molecular Weight:
133.10
MDL number:
UNSPSC Code:
12352209
EC Index Number:
200-291-6
NACRES:
NA.21

Quality Level

form

solid

pH

2.5-3.5 (20 °C, 4 g/L in H2O)

mp

269-271 °C (decomposition)

solubility

4 g/L

bulk density

430 kg/m3

storage temp.

2-30°C

InChI

1S/C4H7NO4/c5-2(4(8)9)1-3(6)7/h2H,1,5H2,(H,6,7)(H,8,9)

InChI key

CKLJMWTZIZZHCS-UHFFFAOYSA-N

Analysis Note

Assay (alkalimetric, calculated on dried substance): ≥ 99.0 %
Identity (IR-spectrum): passes test
Appearance: white to almost white crystalline powder or colorless crystals
Appearance of solution (0.5 mol/l, hydrochloric acid 1 mol/l): clear and colorless
Spec. rotation (α 20/D, 50 g/l, hydrochloric acid 1 mol/l, calc. on dried substance): +25.3 to +26.3
UV-Absorption (260 nm, 0.5 mol/l, 1 cm, hydrochloric acid 1 mol/l): ≤ 0.25
UV-Absorption (280 nm, 0.5 mol/l, 1 cm, hydrochloric acid 1 mol/l): ≤ 0.20
Chloride (Cl): ≤ 0.02 %
Sulfate (SO₄): ≤ 0.01 %
Heavy metals (as Pb): ≤ 0.001 %
As (Arsenic): ≤ 0.0005 %
Ca (Calcium): ≤ 0.001 %
Co (Cobalt): ≤ 0.0005 %
Fe (Iron): ≤ 0.0005 %
K (Potassium): ≤ 0.0005 %
Mg (Magnesium): ≤ 0.0005 %
Na (Sodium): ≤ 0.005 %
NH₄ (Ammonium): ≤ 0.01 %
Zn (Zinc): ≤ 0.0005 %
Foreign amino acids: ≤ 0.3 %
Other ninhydrine positive substances: ≤ 0.1 %
Loss on drying (105 °C; 3 h): ≤ 0.3 %

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service