Skip to Content
MilliporeSigma

09-274

Anti-PEAK1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

NKF3 kinase family member, Sugen kinase 269

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
4
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PEAK1(79834)

General description

250 kDa
PEAK1 belongs to the protein kinase superfamily. PEAK1 is also called Tyrosine-protein kinase SgK269 and Sugen kinase 269. It contains a divergent and possibly inactive eukaryotic protein kinase domain. It has one human paralog, known as SgK223 or pragmin. These two constitute the NKF3 subfamily of protein kinases, found in vertebrates and echinoderms.

Immunogen

Epitope: 1511-1660 aa
Recombinant Protein, aa: 1511-1660. Homology: 100% human and chimp; 96% pig and horse; 90% mouse; 88% rat

Application

This Anti-PEAK1 Antibody is validated for use in WB for the detection of PEAK1.

Biochem/physiol Actions

Other species have not been tested.

Analysis Note

Control
FG Nonstim /Nonstrvd lysate
Routinely evaluated by Western Blot on FG Nonstim /Nonstrvd lysate.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library