Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ELISA, IHC, RIA
Citations:
2
biological source
rabbit
Quality Level
conjugate
unconjugated
antibody form
serum
antibody product type
primary antibodies
clone
polyclonal
species reactivity (predicted by homology)
all
manufacturer/tradename
Upstate®
technique(s)
ELISA: suitable, immunohistochemistry: suitable, radioimmunoassay: suitable
isotype
IgG
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... PRKG1(5592)
Immunogen
2 Monosuccinyl cyclic GMP conjugated to KLH
Application
Anti-cGMP (cyclic GMP) Antibody detects level of cGMP (cyclic GMP) & has been published & validated for use in ELISA, IH, RIA.
Research Category
Signaling
Signaling
Research Sub Category
GPCR, cAMP/cGMP & Calcium Signaling
GPCR, cAMP/cGMP & Calcium Signaling
Biochem/physiol Actions
Recognizes cGMP. Very little cross reactivity to cAMP based on ELISA.
Physical form
Serum
Preparation Note
2 years at -20°C from date of shipment
Analysis Note
Routinely evaluated by ELISA
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
10 - Combustible liquids
wgk
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Chongyang Zhang et al.
Journal of the American Heart Association, 11(2), e023053-e023053 (2022-01-06)
Background cGMP-hydrolyzing phosphodiesterase type 5 (PDE5) regulates vascular smooth muscle cell (SMC) contraction by antagonizing cGMP-dependent protein kinase I (PKGI)-dependent SMC relaxation. SMC contractile dysfunction is implicated in the pathogenesis of aortic aneurysm. PDE5 inhibitors have been used for treating
Susanne Vogeler et al.
BMC developmental biology, 20(1), 23-23 (2020-11-25)
Nitric oxide (NO) is presumed to be a regulator of metamorphosis in many invertebrate species, and although NO pathways have been comparatively well-investigated in gastropods, annelids and crustaceans, there has been very limited research on the effects of NO on
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 09-101 | 04053252475542 |