Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

07-868

Sigma-Aldrich

Anti-phospho-PKA Catalytic β subunit (Ser338) Antibody

Upstate®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pSer338)

Gene Information

human ... PRKACB(5567)

Specificity

Predicted to cross-react with human based on sequence homology (100% homologous).
Recognizes phospho-PKA Catalytic β subunit (Ser338).

Immunogen

Peptide corresponding to amino acid region encompassing the human, cow, and pig phospho-PKA Catalytic β (Ser338).

Application

Research Category
Signaling
Research Sub Category
Cytoskeletal Signaling
This Anti-phospho-PKA Catalytic β subunit (Ser338) Antibody is validated for use in WB for the detection of phospho-PKA Catalytic β subunit (Ser338).

Quality

Routinely evaluated by immunoblot.

Target description

~40 kDa

Physical form

100 μL of affinity purified rabbit polyclonal IgG in 50% storage buffer (PBS (without Mg2+ and Ca2+), pH 7.3 containing 1.0 mg/mL BSA (IgG, protease free) and 0.05% sodium azide) and 50% glycerol.
ImmunoAffinity Purified

Storage and Stability

Stable for 2 years at -20°C from date of shipment.
Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. Note: Variability in freezer temperatures below -20°C may cause glycerolcontaining solutions to become frozen during storage.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takamasa Ishii et al.
Aging cell, 16(1), 39-51 (2016-09-15)
The etiology of astrocyte dysfunction is not well understood even though neuronal defects have been extensively studied in a variety of neuronal degenerative diseases. Astrocyte defects could be triggered by the oxidative stress that occurs during physiological aging. Here, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service