Skip to Content
MilliporeSigma

07-617

Anti-mGluR1 Antibody

Upstate®, from rabbit

Synonym(s):

Anti-GPRC1A, Anti-MGLU1, Anti-MGLUR1, Anti-PPP1R85, Anti-SCA44, Anti-SCAR13

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
25
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GRM1(2911)

General description

142kDa

Immunogen

peptide corresponding to amino acids 1180-1199 (C-PNVTYASVILRDYKQSSSTL) of rat mGluR1

Application

Anti-mGluR1 Antibody detects level of mGluR1 & has been published & validated for use in WB.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Neurodegenerative Diseases

Biochem/physiol Actions

Predicted to cross-react with human and mouse based on sequence homology
mGluR1

Physical form

0.1M Tris-glycine, pH 7.4, 0.15M NaCl, and 0.05% sodium azide.
Format: Purified
Protein A purified

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

routinely evaluated by immunoblot on mGluR1 in rat brain microsomal preparation

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library