Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

07-425-I

Sigma-Aldrich

Anti-Neurogranin

from rabbit, purified by affinity chromatography

Synonym(s):

Ng, RC3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human

species reactivity (predicted by homology)

goat (based on 100% sequence homology), bovine (based on 100% sequence homology), mouse (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... NRGN(4900)

General description

Neurogranin (UniProt: Q92686; also known as Ng, RC3) is encoded by the NRGN gene (Gene ID: 4900) in human. Neurogranin is abundantly expressed in cerebral cortex, hippocampus, amygdala, and striatum. In neurons, it is found concentrated at dendritic spines. It acts as a third messenger and serves as a substrate for protein kinase C-mediated molecular cascades during synaptic development and remodeling. It contains an IQ domain (aa 26-347) and a collagen-like domain (aa 48-78). The IQ domain mediates its interaction with calmodulin and phosphatidic acid (PA). Neurogranin is intrinsically unstructured, however, upon binding with calmodulin, the IQ domain adopts a helical conformation. Neurogranin is phosphorylated at serine 36 by protein kinase C and following phosphorylation neurogranin is unable to bind either calmodulin or PA, which can lead to decline in memory-associated functions. Higher levels of neurogranin have also been reported in brains of elderly and is considered a factor in memory decline, independent of Alzheimer s disease. (Ref.: Javier Dı´ez-Guerra, F. (2010). Life. 62(8): 597-606; Casaletto, KB et al. (2017). Neurology 89(17); 1782-1788).

Specificity

This rabbit polyclonal antibody sepecifically detects Neurogranin. It targets an epitope with in 12 amino acids from the C-terminal region.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to 12 amino acids from the C-terminal region of human Neurogranin.

Application

Anti-Neurogranin Antibody, Cat. No. 07-425-I, is a highly specific rabbit polyclonal antibody that targets Neurogranin and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected Neurogranin in human and rat brain tissue sections.
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in rat brain tissue lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected Neurogranin in rat brain tissue lysate.

Target description

~15 kDa observed; 7.62 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ashton N Jorgensen et al.
Redox biology, 70, 103085-103085 (2024-02-16)
Endothelial dysfunction and endothelial activation are common early events in vascular diseases and can arise from mitochondrial dysfunction. Neurogranin (Ng) is a 17kD protein well known to regulate intracellular Ca2+-calmodulin (CaM) complex signaling, and its dysfunction is significantly implicated in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service