Skip to Content
MilliporeSigma

07-404

Anti-PRMT1 Antibody

Upstate®, from rabbit

Synonym(s):

Anti-ANM1, Anti-HCP1, Anti-HRMT1L2, Anti-IR1B4

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
68
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PRMT1(3276)

General description

42-44 kDa
The methylation of arginine is mediated by the Protein Arginine Methyl Transferase (PRMT) family of enzymes. These enzymes transfer the methyl group from S-adenosyl-Lmethionine to the guanidino nitrogen atoms of an arginine residue. Arginine methylation was found to be an important modification in signal transduction, transcription, RNA transport, and splicing. PRMT1, also known as HRMT1L2 and IR1B4, was isolated through its interaction with BTG1 and TIS21, proteins that are important in cell quiescence. PRMT1 is a 361 amino acid protein; its splicing variants differ at the N-terminus, giving rise to three isoforms of 343, 361, and 347 amino acids (variants 1, 2, and 3 respectively). Proteins involved in the metabolism of RNA are substrates for PRMT1, among them Sam68.

Immunogen

Peptide corresponding to residues 298-318 of human protein arginine methyltransferase 1.

Application

Anti-PRMT1 Antibody is a high quality Rabbit Polyclonal Antibody for the detection of PRMT1 & has been validated in WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Biochem/physiol Actions

PRMT1 doublet

Physical form

0.1M Tris-glycine, pH 7.4, 0.15M NaCl,0.05% sodium azide before the addition of glycerol to 30%
Format: Purified
Protein A purified

Preparation Note

2 years at -20°C

Analysis Note

Control
Positive Antigen Control: Catalog #12-303, Jurkat cell lysate.
routinely evaluated by immunoblot in lysates from Jurkat and 3T3/A31, NIH 3T3, HeLa, and mouse ES cells

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library