Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

07-360-S

Sigma-Aldrich

Anti-acetyl-Histone H3 (Lys27) Antibody, Trial Size

serum, Upstate®

Synonym(s):

H3K27Ac, Histone H3 (acetyl K27)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

vertebrates, Saccharomyces cerevisiae, human

manufacturer/tradename

Upstate®

technique(s)

ChIP: suitable (ChIP-seq)
dot blot: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

acetylation (Lys27)

Gene Information

human ... HIST1H3F(8968)

General description

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure.



The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.

Specificity

Recognizes Histone H3 acetylated on lysine 27.

Immunogen

Epitope: Lys27
Synthetic peptide corresponding to the sequence AR(acK)SA, in which acK corresponds to acetyl lysine 27 of yeast histone H3.

Application

ChIP-seq Analysis: Chromatin immunoprecipitation was performed using the Magna ChIP HiSens kit (cat# 17-10460), 2 µl of anti-acetyl-Histone H3 (Lys27) antibody (cat# 07-360-S), 20 µL Protein A/G beads, and 1e6 crosslinked HeLa cell chromatin followed by DNA purification using magnetic beads. Libraries were prepared from Input and ChIP DNA samples using standard protocols with Illumina barcoded adapters, and analyzed on Illumina HiSeq instrument. An excess of fourteen million reads from FastQ files were mapped using Bowtie (http://bowtie-bio.sourceforge.net/manual.shtml) following TagDust (http://genome.gsc.riken.jp/osc/english/dataresource/) tag removal. Peaks were identified using MACS (http://luelab.dfci.harvard.edu/MACS/), with peaks and reads visualized as a custom track in UCSC Genome Browser (http://genome.ucsc.edu) from BigWig and BED files. The highest 25% of peaks identified in the 07-360 dataset showed 96% overlap with peaks identified in the ENCODE H3K27Ac BROAD Histone track for HeLa S3.
Anti-acetyl-Histone H3 (Lys27) Antibody is a Rabbit Polyclonal Antibody for detection of Histone H3 acetylated at lysine 27 (H3K27ac). This antibody has dot blot verified specificity and has been validated in ChIP, WB & Mplex.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Quality

routinely evaluated by immunoblot on acid extracts from sodium butyrate treated HeLa cells

Target description

17 kDa

Physical form

Immunoaffinity Purified immunoglobulin in 0.2M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide, and 5mg/ml BSA before the addition of glycerol to 30%
Unpurified

Storage and Stability

Maintain for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Acid extracted proteins from HeLa cells treated with sodium butyrate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lin Yu et al.
Heliyon, 10(4), e25725-e25725 (2024-02-23)
A super-enhancer (SE) is a huge cluster of multiple enhancers that control the key genes for cell identity and function. The rise of advanced chromatin immunoprecipitation sequencing (ChIP-seq) technology such as Cleavage Under Targets and Tagmentation (CUT&Tag) allows more SEs
Hazem E Hassan et al.
Epigenetics, 11(10), 740-749 (2016-09-03)
Curcumin and its analogs exhibited antileukemic activity either as single agent or in combination therapy. Dimethoxycurcumin (DMC) is a more metabolically stable curcumin analog that was shown to induce the expression of promoter-methylated genes without reversing DNA methylation. Accordingly, co-treatment

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service