Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

07-108

Sigma-Aldrich

Anti-Histone H4 Antibody

Upstate®, from rabbit

Synonym(s):

H4 histone family, member A, histone 1, H4a, histone cluster 1, H4a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

Xenopus, mouse, avian, chicken, human, bovine

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

bovine ... H4C1(617905)
frog ... H4C1(549623)
human ... H4C1(8359)
mouse ... H4C1(326619)

General description

Histone H4 is one of the 5 main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N terminal tail H4 is involved with the structure of the nucleosomes of the ′beads on a string′ structure.

Specificity

Broad species cross-reactivity expected.
Recognizes Histone H4, Mr 10 kDa.

Immunogen

Calf thymus histone H4.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
Use Anti-Histone H4 Antibody (Rabbit Polyclonal Antibody) validated in WB to detect Histone H4 also known as Histone H4.

Quality

Routinely evaluated by western blot on acid extracts of HeLa cells treated with sodium butyrate.

Western Blot Analysis:
0.1-0.5 μg/mL of this lot detected Histone H4 in acid extracts of HeLa cells treated with or without 5 mM sodium butyrate. This lot did not cross react with Histone H2A, Histone H2B or Histone H3.

Target description

10 kDa

Physical form

Format: Purified
Protein A chromatography
Purified rabbit polyclonal IgG in buffer containing 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% Sodium Azide.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 6 months after date of receipt.

Analysis Note

Control
HeLa cell extract.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regulation of chemokine gene expression by hypoxia via cooperative activation of NF-kappaB and histone deacetylase.
Olga Safronova,Sireerat Pluemsampant,Ken-ichi Nakahama,Ikuo Morita
The International Journal of Biochemistry & Cell Biology null
Ingeborg M M van Leeuwen et al.
Molecular cancer therapeutics, 12(4), 471-480 (2013-02-19)
Acetylation of C-terminal lysine residues in the p53 tumor suppressor is associated with increased stability and transcription factor activity. The function, protein level, and acetylation of p53 are downregulated by mdm2, which in its turn is inhibited by the p14(ARF)
S L Gilbert et al.
The Journal of biological chemistry, 275(47), 36491-36494 (2000-09-28)
Microscopy studies have shown that XIST RNA colocalizes with the inactive X chromosome (Xi). However, the molecular basis for this colocalization is unknown. Here we provide two lines of evidence from chromatin immunoprecipitation experiments that XIST RNA physically associates with
Setdb1-mediated histone H3K9 hypermethylation in neurons worsens the neurological phenotype of Mecp2-deficient mice.
Jiang, Y; Matevossian, A; Guo, Y; Akbarian, S
Neuropharmacology null
Dongcheng Wu et al.
The Journal of biological chemistry, 277(14), 12001-12008 (2002-01-29)
Chromatin structure is influenced by histone modification, and this may help direct chromatin behavior to facilitate transcription, DNA replication, and DNA repair. Chromatin condensation and DNA fragmentation are the classic nuclear features but remain poorly characterized. It is highly probable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service