Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

06-556

Sigma-Aldrich

Anti-Kip2 Antibody

Upstate®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, bovine

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... CDKN1C(1028)

Specificity

Kip2

Immunogen

15 amino acid synthetic peptide (CGVGSVEQTPRKRLR) corresponding to amino acids 303-316 of human Kip2

Application

Detect Kip2 with Anti-Kip2 Antibody (Rabbit Polyclonal Antibody), that has been shown to work in IP & WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair

Quality

routinely evaluated by immunoblot on HeLa nuclear extract or bovine brain nuclear extract

Target description

57kDa

Physical form

0.1M Tris-glycine, pH 7.4, 0.15M NaCl and 0.05% sodium azide
Format: Purified
Protein A chromatography

Storage and Stability

2 years at -20°C

Analysis Note

Control
Positive Antigen Control: Catalog #12-309, Hela cell nuclear extract. Add an equal volume of Laemmli reducing sample buffer to 10 μL of extract and boil for 5 minutes to reduce the preparation. Load 20 μg of reduced extract per lane for minigels.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steffen Heeg et al.
Cancer science, 102(2), 351-360 (2010-12-16)
Telomerase plays an important role during immortalization and malignant transformation as crucial steps in the development of human cancer. In a cellular model of oral-esophageal carcinogenesis, recapitulating the human disease, immortalization occurred independent of the activation of telomerase but through
Cyclin-dependent kinase inhibitor p57KIP2 in soft tissue sarcomas and Wilms'tumors.
Orlow, I, et al.
Cancer Research, 56, 1219-1221 (1996)
S Matsuoka et al.
Genes & development, 9(6), 650-662 (1995-03-15)
Cyclin-dependent kinases (Cdks) are positive regulators of cell proliferation, whereas Cdk inhibitors (CKIs) inhibit proliferation. We describe a new CKI, p57KIP2, which is related to p21CIP1 and p27KIP1. p57KIP2 is a potent, tight-binding inhibitor of several G1 cyclin/Cdk complexes, and
M Jeanblanc et al.
Oncogene, 31(25), 3072-3085 (2011-10-25)
We developed a clonal WI-38hTERT/GFP-RAF1-ER immortal cell line to study RAF-induced senescence of human fibroblasts. Activation of the GFP-RAF1-ER kinase by addition of 4-hydroxy-tamoxifen led to a robust induction of senescence within one population doubling, accompanied by the assembly of
Dilli Ram Bhandari et al.
Journal of cellular and molecular medicine, 15(7), 1603-1614 (2010-08-19)
Myelocytomatosis oncogene (c-MYC) is a well-known nuclear oncoprotein having multiple functions in cell proliferation, apoptosis and cellular transformation. Chromosomal modification is also important to the differentiation and growth of stem cells. Histone deacethylase (HDAC) and polycomb group (PcG) family genes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service