Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

06-501

Sigma-Aldrich

Anti-STAT1 Antibody, CT

Upstate®, from rabbit

Synonym(s):

Anti-CANDF7, Anti-IMD31A, Anti-IMD31B, Anti-IMD31C, Anti-ISGF-3, Anti-STAT91

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

manufacturer/tradename

Upstate®

technique(s)

electrophoretic mobility shift assay: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... STAT1(6772)

General description

STAT proteins (Signal Transduction and Activators of Transcription) are latent cytoplasmic transcription factors that have the dual function of signal transduction and activation of transcription. STATs are activated by tyrosine phosphorylation in response to different ligands, after which they translocate to the cell nucleus. The N-terminal region is highly homologous among the STAT proteins and surrounds a completely conserved arginine residue. STATs are a part of the JAK-STAT signaling pathway – a major pathway of the immune system. All cytokines transduce critical signals through this pathway.

STAT 1, is activated by a number of different ligands, including IFNalpha, IFNgamma, EGF, PDGF and IL6. Phosphorylation of tyrosine 701 is required for STAT 1 dissociation from IFNGR1, homodimerization, and nuclear translocation. Tyrosine 701 phosphorylation impairment results in loss of STAT 1 functions.

Specificity

Recognizes STAT1.

Immunogen

GST fusion-protein containing the carboxy terminal 36 amino acids of human STAT1α

Application

Detect STAT1 also known as Signal Transducer & Activator of Transcription 1 with Anti-STAT1 Antibody, CT (Rabbit Polyclonal Antibody), that has been demonstrated to work in EMSA, IP & WB.

Quality

routinely evaluated by immunoblot on RIPA lysates from human Jurkat cells, mouse CTLL or rat L6 cells

Target description

91 kDa

Physical form

Format: Purified

Analysis Note

Control
Positive Antigen Control: Catalog #12-303, Jurkat cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

IL-4 suppresses the responses to TLR7 and TLR9 stimulation and increases the permissiveness to retroviral infection of murine conventional dendritic cells.
Sriram, U; Xu, J; Chain, RW; Varghese, L; Chakhtoura, M; Bennett, HL; Zoltick, PW; Gallucci, S
Testing null
Nipah virus V protein evades alpha and gamma interferons by preventing STAT1 and STAT2 activation and nuclear accumulation.
Rodriguez, JJ; Parisien, JP; Horvath, CM
Journal of virology null
Saehyung Lee et al.
Frontiers in pharmacology, 9, 1568-1568 (2019-02-09)
The glycoengineering approach is used to improve biophysical properties of protein-based drugs, but its direct impact on binding affinity and kinetic properties for the glycoengineered protein and its binding partner interaction is unclear. Type I interferon (IFN) receptors, composed of
Thrombopoietin signal transduction in purified murine megakaryocytes.
Drachman, J G, et al.
Blood, 89, 483-492 (1997)
A F Wilks et al.
BioEssays : news and reviews in molecular, cellular and developmental biology, 16(5), 313-320 (1994-05-01)
Cytokine receptors fall into two basic classes: those with their own intrinsic protein tyrosine kinase (PTK) domain, and those lacking a PTK domain. Nonetheless, PTK activity plays a fundamental role in the signal transduction processes lying downstream of both classes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service