Skip to Content
MilliporeSigma

06-1398

Anti-CDK5RAP2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

CDK5 regulatory subunit-associated protein 2, CDK5 activator-binding protein C48, Centrosome-associated protein 215

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, WB
Citations:
27
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunocytochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CDK5RAP2(55755)

General description

Also known as Cep215, CDK5RAP2 is involved in a variety of cellular processes. It serves as a potential regulator of CDK5 activity through its interaction with CDK5R1. It acts as a negative regulator of centriole disengagement, which maintains centriole engagement and cohesion. It is involved in the regulation of mitotic spindle orientation. Additionally, this protein mediates spindle checkpoint activation by serving as a transcriptional regulator of the MAD2 and BUBR1 promoter. In conjuction with MAPRE1, CDK5RAP2 promotes microtubule polymerization, bundle formation, and growth at the plus ends. It also plays a role in DNA damage-induced G2 cell cycle checkpoint. CDK5RAP2 is known to be mutated in primary microcephaly, a neuodevelopmental disorder that is characterized by a decrease in brain size.
~220 kDa observed. Uniprot.org states four isoforms produced by alternative splicing, which may be observed at a molecular weight of ~216 kDa, ~211 kDa, ~210 kDa, and ~206 kDa in some cell lysates.

Immunogen

MBP-tagged recombinant protein corresponding to human CDK5RAP2.

Application

Immunocytochemistry Analysis: A representative lot from an independent laboratory detected CDK5RAP2 in RPE1 cells. (Barr, A.R., et al. (2010). J Cell Biol. 189(1):23-39.)
Use Anti-CDK5RAP2 Antibody (rabbit polyclonal antibody) validated in WB, ICC to detect CDK5RAP2 also known as CDK5 regulatory subunit-associated protein 2, CDK5 activator-binding protein C48, Centrosome-associated protein 215.

Biochem/physiol Actions

Other Homologies: Rat (83% sequence homology) and Mouse (82% sequence homology).

Analysis Note

Evaluated by Western Blot in 10 µg of HeLa cell lysate.

Western Blot Analysis: A 1:2,500 dilution of this antibody dectected CDK5RAP2 in 10µg of HeLa cell lysate.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library