Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

05-903

Sigma-Aldrich

Anti-GSK3α/β Antibody, rabbit monoclonal

Upstate®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

monoclonal

species reactivity

human, mouse

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GSK3A(2931)

Specificity

GSK3 alpha and beta
Predicted to cross-react with rat based on sequence homology

Immunogen

N-terminal 6His-tagged recombinant full length human GSK3β

Application

Anti-GSK3α/β Antibody is an antibody against GSK3α/β for use in IP & WB.

Quality

routinely evaluated by immunoblot on lysates from HEK293 cells

Target description

Mr 46kDa (beta) and 51kDa (alpha)

Physical form

Format: Purified

Analysis Note

Control
Positive Antigen Control: Catalog #12-303, Jurkat cell lysate.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lithium and GSK-3: one inhibitor, two inhibitory actions, multiple outcomes.
Jope, Richard S
Trends in Pharmacological Sciences, 24, 441-443 (2003)
Constitutive mTORC1 activation by a herpesvirus Akt surrogate stimulates mRNA translation and viral replication.
Chuluunbaatar, U; Roller, R; Feldman, ME; Brown, S; Shokat, KM; Mohr, I
Genes & Development null
Yasuhiro Nishino et al.
Circulation research, 103(3), 307-314 (2008-06-28)
The inactivation of glycogen synthase kinase-3beta (GSK-3beta) is proposed as the event integrating protective pathways initiated by preconditioning and other interventions. The inactivation of GSK-3 is thought to decrease the probability of opening of the mitochondrial permeability transition pore. The
Jen Q Pan et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 36(7), 1397-1411 (2011-03-11)
Bipolar disorder (BP) is a debilitating psychiatric disorder, affecting ∼2% of the worldwide population, for which the etiological basis, pathogenesis, and neurocircuitry remain poorly understood. Individuals with BP suffer from recurrent episodes of mania and depression, which are commonly treated
The neuron-specific isoform of glycogen synthase kinase-3beta is required for axon growth.
Casta?o, Zafira, et al.
Journal of Neurochemistry, 113, 117-130 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service