Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

05-882

Sigma-Aldrich

Anti-Parkin Antibody, clone PRK8

ascites fluid, clone PRK8, Upstate®

Synonym(s):

Anti-AR-JP, Anti-LPRS2, Anti-PARK2, Anti-PDJ

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

PRK8, monoclonal

species reactivity

human, mouse

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... PARK2(5071)

Specificity

Parkin

Immunogen

Full length recombinant human Parkin. Epitope has been mapped to the second ring domain (aa 399-465)

Application

Anti-Parkin Antibody, clone PRK8 detects level of Parkin & has been published & validated for use in IP, WB, IC, IH.

Quality

routinely evaluated by immunoblot on RIPA lysates from HEK293 cells

Target description

52kDa May be observed as a doublet at 52 and 46kDa

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genotype-phenotype correlation: familial Parkinson disease.
Mori, Hideo, et al.
Neuropathology : Official Journal of the Japanese Society of Neuropathology, 23, 90-94 (2003)
Nobutaka Hattori et al.
Lancet (London, England), 364(9435), 722-724 (2004-08-25)
The cause and pathogenesis of Parkinson's disease remain unknown; mitochondrial dysfunction, oxidative damage, environmental factors, and genetic predisposition might all be involved. Identification of the causative genes for familial Parkinson's diseases allow study of the pathogenesis of the disease at
Christian Pennanen et al.
Journal of cell science, 127(Pt 12), 2659-2671 (2014-04-30)
Cardiomyocyte hypertrophy has been associated with diminished mitochondrial metabolism. Mitochondria are crucial organelles for the production of ATP, and their morphology and function are regulated by the dynamic processes of fusion and fission. The relationship between mitochondrial dynamics and cardiomyocyte
Keiji Tanaka et al.
Biochimica et biophysica acta, 1695(1-3), 235-247 (2004-12-02)
The ubiquitin-proteasome system (UPS) is important for intracellular proteolysis, and is responsible for a diverse array of biologically important cellular processes, such as cell-cycle progression, signaling cascades and developmental programs. This system is also involved in the protein quality control
Molecular pathways of neurodegeneration in Parkinson's disease.
Dawson, Ted M and Dawson, Valina L
Science (New York, N.Y.), 302, 819-822 (2003)

Articles

Autophagy is a regulated process involved in cell growth, development, and recycling of cytoplasmic components in cells.

Autophagy is a regulated process involved in cell growth, development, and recycling of cytoplasmic components in cells.

Autophagy is a regulated process involved in cell growth, development, and recycling of cytoplasmic components in cells.

Autophagy is a regulated process involved in cell growth, development, and recycling of cytoplasmic components in cells.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service