Skip to Content
MilliporeSigma

Skip To

05-689

Anti-HP1α Antibody, clone15.19s2

clone 15.19s2, Upstate®, from mouse

Synonym(s):

Antigen p25, HP1 alpha, HP1Hs alpha, Heterochromatin protein 1 homolog alpha, chromobox homolog 5 (Drosophila HP1 alpha), chromobox homolog 5 (HP1 alpha homolog, Drosophila), heterochromatin protein 1-alpha

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
15.19s2, monoclonal
Application:
ChIP, ICC, IHC, IP, WB
Citations:
94
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

15.19s2, monoclonal

species reactivity

mouse, vertebrates, human

manufacturer/tradename

Upstate®

technique(s)

ChIP: suitable, immunocytochemistry: suitable, immunohistochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CBX5(23468)

General description

29 kDa
Heterochromatin protein 1 (HP1) is a family of heterochromatic adaptor molecules involved in both gene silencing and higher order chromatin structure. All three HP1 family members (α, β, and γ) are primarily associated with centromeric heterochromatin; however, HP1β and γ also localize to euchromatic sites in the genome. HP1 proteins are approximately 25 kDa in size and each contains a conserved amino-terminal chromodomain, followed by a variable hinge region and a conserved carboxy-terminal chromoshadow domain. The chromodomain facilitates binding to histone H3 tri-methylated on Lys9, a histone "mark" closely associated with centromeric heterochromatin. The variable hinge region binds both RNA and DNA in a sequence-independent manner. The chromoshadow domain mediates the dimerization of HP1 proteins, in addition to binding multiple proteins implicated in gene silencing and heterochromatin formation, including the SUV39H histone methyltransferase, the DNMT1 and DNMT3a DNA methyltransferases and the p150 subunit of chromatin-assembly factor-1 (CAF1). In addition to contributing to heterochromatin formation and propagation, HP1 and SUV39H are also found complexed with retinoblastoma (Rb) and E2F6 proteins, both of which function to repress euchromatic gene transcription in quiescent cells. HP1 proteins are subject to multiple types of post-translational modifications, including phosphorylation, acetylation, methylation, ubiquitination and sumoylation, suggesting multiple means of regulation.

Immunogen

GST fusion protein corresponding to residues 2-191 of human HP1α protein.

Application

Anti-HP1α Antibody, clone15.19s2 is a Mouse Monoclonal Antibody for detection of HP1α also known as HP1Hs alpha, Heterochromatin protein 1 homolog alpha & has been validated in ChIP, ICC, IHC, IP & WB.
Immunohistochemistry:
A previous lot of this antibody has been reported by an independent laboratory to detect HP1a in NIH/3T3 nuclei (Ayyanathan, K., 2003).

Immunoprecipitation/Chromatin Immunoprecipitation:
A previous lot of this antibody has been reported by an independent laboratory to immunoprecipitate HP1α from nuclear extracts and formalin-cross-linked chromatin (Ayyanathan, K., 2003).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

RNA Binding Protein (RBP)

Biochem/physiol Actions

This antibody recognizes HP1α

Physical form

Format: Purified
Protein G Chromatography
Purified mouse monoclonal IgG1 in buffer containing 0.1M Tris-Glycine, 0.15M NaCl, 0.05% Sodium Azide, pH 7.4.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Handling Recommendations:


Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
HeLa, NIH/3T3 and COS cell lysates.
Routinely evaluated by western blot in acid-extracted HeLa cell lysates.

Western Blot Analysis:
0.5-2 µg/mL of this lot detected HP1α protein in acid-extracted HeLa cell lysates.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Due to license agreement restrictions, this product cannot be purchased for resale.
Replaces: MABE479

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters