Skip to Content
MilliporeSigma

05-677

Anti-phospho-Src (Tyr416) Antibody, clone 9A6

clone 9A6, Upstate®, from mouse

Synonym(s):

proto-oncogene tyrosine-protein kinase SRC, protooncogene SRC, Rous sarcoma, tyrosine kinase pp60c-src, tyrosine-protein kinase SRC-1, v-src avian sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog, v-src sarcoma (Schmidt-Ruppin A-2) viral oncogene homo

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
9A6, monoclonal
Species reactivity:
mouse, human
Application:
IP, WB
Citations:
38
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9A6, monoclonal

species reactivity

mouse, human

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable, western blot: suitable

isotype

IgG3

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pTyr416)

Gene Information

human ... SRC(6714)

General description

60 kDa
Specific src isoform is unknown
Src (also known as pp60src) is a non receptor Tyrosine Kinase involved in signal transduction in many biological systems and implicated in the development of human tumors. There are two critical phosphorylation sites of tyrosine on Src, tyrosine 418 and tyrosine 529 (referring to human Src sequence). The tyrosine 418 is located in the catalytic domain and is one of the autophosphorylation sites. Full catalytic activity of Src requires phosphorylation of tyrosine 418. This region of Src is also highly conserved in all of the related Src-family kinases, and thus prior immunoprecipitation may be required to identify which Src family member is being activated.

Immunogen

KLH-conjugated, synthetic peptide containing phosphotyrosine in the sequence context corresponding to tyrosine 416 of avian Src. Clone 9A6.

Application

Detect phospho-Src (Tyr416) using this Anti-phospho-Src (Tyr416) Antibody, clone 9A6 validated for use in WB & IP.
Immunoprecipitation/Immunoblot Analysis: 4 μg/mL of a previous lot detected phospho-Src immunoprecipitated from 3T3 cell lysates with 10 μg of anti-Src (Catalog # 05-184), but not after phosphatase treatment with YOP.
Research Category
Signaling
Research Sub Category
Cytoskeletal Signaling

Biochem/physiol Actions

Recognizes phospho-Src, Mr 60kDa. The antibody does not crossreact with the nonphosphorylated form of Src or with unrelated phosphorylation sites. Predicted to recognize all Src family members phosphorylated at the tyrosine corresponding to Tyr416 of avian Src.

Physical form

Format: Purified
Purified mouse monoclonal IgG3 lyophilized from 1 mL 2x PBS / 0.09 % Na-azide / PEG and Sucrose. Reconstitute with 1 mL H2O (15 min, RT). Lyophilized. Store at -20ºC.
Thiophilic adsoption and size exclusion chromatography

Preparation Note

Stable for 1 year at from date of receipt.

Analysis Note

Control
Chick embryo fibroblast (CEF) cells expressing human wild-type Src protein
Evaluated by immunoprecipitating Src from 3T3 cell lysates with anti-Src (Catalog #05-184), but not after phosphatase treatment with YOP.

Src Autophosphorylation/Western Blotting Analysis: 0.5-2 μg/mL of this antibody strongly detected recombinant Src (Catalog # 14-326) after incubation with Mg/Mn/ATP. Phospho-specificity was confirmed by dephosphorylation of the Src protein with λ-phosphatase.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

11 - Combustible Solids

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library