Skip to Content
MilliporeSigma

05-175

Anti-Myb Antibody, clone 1-1

clone 1-1, Upstate®, from mouse

Synonym(s):

Avian myeloblastosis viral (v-myb) oncogene homolog 2 c-myb protein (140 AA), v-myb avian myeloblastosis viral oncogene homolog, v-myb avian myeloblastosis viral oncogene homolog, v-myb myeloblastosis viral oncogene homolog, v-myb myeloblastosis viral on

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
1-1, monoclonal
Application:
IHC, IP, WB
Citations:
78
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1-1, monoclonal

species reactivity

mouse, human

manufacturer/tradename

Upstate®

technique(s)

immunohistochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... MYB(4602)

General description

80 kDa
Myb is a transcriptional activator. It is a DNA-binding protein that specifically recognizes the sequence 5′-YAAC[GT]G-3′. Myb plays an important role in the control of proliferation and differentiation of hematopoietic progenitor cells.

Immunogen

Recombinant murine Myb purified by SDS-PAGE (Nicot, C., 2001) Clone 1-1.

Application

Anti-Myb Antibody, clone 1-1 is a Mouse Monoclonal Antibody for detection of Myb also known as Avian myeloblastosis viral (v-myb) oncogene homolog 2 c-myb protein (140 AA) & has been validated in IHC, IP & WB.
Immunoprecipitation:
4 μg of previous lot immunoprecipitated Myb from Jurkat cell lysate.

Immunohistochemistry:
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Biochem/physiol Actions

This antibody recognizes Myb, Mr 80 kDa, containing C-terminal amino acids from 401-636 (Salomoni, P., 1997).

Physical form

Ammonium sulfate precipitation, gel filtration, and ion-exchange chromatography.
Format: Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-glycine, pH 7.4, 0.15 M NaCl and 0.05% sodium azide. Frozen solution.

Preparation Note

Stable for 1 year at -20°C from date of receipt. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Positive Antigen Control: Catalog #12-303, Jurkat cell lysate.
Routinely evaluated on RIPA lysate from Jurkat cells.

Western Blot Analysis:
0.5-2 μg/mL of this lot detected Myb in 20 μg of RIPA lysate from Jurkat cells.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library