Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

05-1352

Sigma-Aldrich

Anti-Histone H2B Antibody, clone 5HH2-2A8

ascites fluid, clone 5HH2-2A8, from mouse

Synonym(s):

H2B, Histone H2B, histone 2, H2be, histone cluster 2, H2be, H2B histone family, member Q

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

5HH2-2A8, monoclonal

species reactivity

mouse, human, rat

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... H2BC21(8349)
mouse ... H2Bc21(319190)

General description

Histones are basic nuclear proteins that are responsible for the nucleosome structure of chromatin in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which DNA is wrapped in repeating units, called nucleosomes. Histone H2B is one of the main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N terminal tail H2B is involved with the structure of the nucleosomes of the ′beads on a string′ structure.

Specificity

Antibody recognizes Histone H2B near the C-terminus.

Immunogen

Epitope: C-terminus
Ovalbumin-conjugated synthetic linear peptide corresponding to the C-terminus of Histone H2B.

Application

Anti-Histone H2B Antibody, clone 5HH2-2A8 is a Mouse Monoclonal Antibody for detection of Histone H2B also known as Histone H2B, histone 2 H2be & has been validated in WB, ICC.
Immunocytochemistry:
A 1:500 dilution of a representative lot of this antibody detected Histone H2B in A431, HeLa and NIH/3T3 cells.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Quality

Evaluated by western blot in HepG2 cell lysate.

Western Blot Analysis:
A 1:1,000-1:2,000 dilution of this antibody detected Histone H2B in 10 µg of HepG2 cell lysate.

Target description

~15 kDa

Linkage

Replaces: MABE15

Physical form

Mouse monoclonal IgG1κ ascites with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
HepG2 cell lysate, L6 cell lysate, Mouse Brain Lysate

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

USP7 cooperates with SCML2 to regulate the activity of PRC1.
Lecona, E; Narendra, V; Reinberg, D
Molecular and cellular biology null
Usha P Andley et al.
BMC research notes, 13(1), 315-315 (2020-07-04)
Understanding the mechanisms of cataract formation is important for age-related and hereditary cataracts caused by mutations in lens protein genes. Lens proteins of the crystallin gene families α-, β-, and γ-crystallin are the most abundant proteins in the lens. Single
Louis Legoff et al.
Environment international, 152, 106472-106472 (2021-03-13)
Chlordecone (CD), also known as Kepone, is an organochlorine insecticide that has been used in banana crops in the French West Indies. Due to long-term contamination of soils and water, the population is still exposed to CD. Exposure to CD
Valery F Thompson et al.
Biochemistry, 57(51), 7021-7032 (2018-11-30)
Purified recombinant FUsed in Sarcoma (FUS) assembles into an oligomeric state in an RNA-dependent manner to form large condensates. FUS condensates bind and concentrate the C-terminal domain of RNA polymerase II (RNA Pol II). We asked whether a granule in
Yang Lim et al.
Science (New York, N.Y.), 381(6664), eadi3448-eadi3448 (2023-08-17)
CDC45-MCM2-7-GINS (CMG) helicase assembly is the central event in eukaryotic replication initiation. In yeast, a multi-subunit "pre-loading complex" (pre-LC) accompanies GINS to chromatin-bound MCM2-7, leading to CMG formation. Here, we report that DONSON, a metazoan protein mutated in microcephalic primordial

Articles

Loading controls in western blotting application.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service