Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

05-1321

Sigma-Aldrich

Anti-BMI1 Antibody, clone AF27

clone AF27, from mouse

Synonym(s):

B lymphoma Mo-MLV insertion region 1 homolog, B lymphoma Mo-MLV insertion region 1 homolog (mouse), BMI1 polycomb ring finger oncogene, Polycomb group RING finger protein 4, RING finger protein 51, murine leukemia viral (bmi-1) oncogene homolog, oncogene

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

AF27, monoclonal

species reactivity

human

technique(s)

ChIP: suitable
immunoprecipitation (IP): suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BMI1(648)

General description

The BMI1 protein is a member of the polycomb group (PcG) proteins. PcG proteins form multiprotein complexes that function as transcriptional repressors. Several lines of evidence implicate BMI1 in tumorigenesis. BMI1 cooperates with c-Myc in the generation of lymphomas in double transgenic mice. BMI1 blocks senescence and immortalizes mouse embryo fibroblasts, although not human fibroblasts, and in combination with an activated H-ras gene, leads to neoplastic transformation. The BMI1 gene is amplified in certain mantle cell lymphomas and is overexpressed in a subset of non-small cell lung cancer, colorectal carcinomas, multiple myelomas and medulloblastomas, but not glioblastomas. The transformation of human hematopoietic progenitor cells by the oncogenic fusion protein E2a-Pbx1 requires BMI1.

Specificity

This antibody recognizes BMI1.

Immunogen

MBP-tagged recombinant protein corres-ponding to full length human BMI1.

Application

Anti-BMI1 Antibody, clone AF27 is a mouse monoclonal antibody for detection of BMI1 also known as BMI1 polycomb ring finger oncogene, murine leukemia viral (bmi-1) oncogene homolog & has been validated in IP, ChIP.
Chromatin Immunoprecipitation (ChIP):
A previous lot was used by an independent laboratory in ChIP. (Bracken, A.P., et al., 2007).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

Quality

Evaluated by Immunoprecipitation in MEF1 cell lysate.
Immunoprecipitation Analysis: 10 μg/mL of this antibody immunoprecipitated BMI1 from 100 μg of MEF1 cell lysate.

Target description

~35 kDa

Physical form

Anti-BMI1, clone AF27
Format: Purified
Purified mouse monoclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable at 2-8°C for 1 year from date of receipt.

Analysis Note

Control
MEF1 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hsan-au Wu et al.
The Journal of biological chemistry, 288(51), 36398-36408 (2013-11-07)
Cbx7 is one of five mammalian orthologs of the Drosophila Polycomb. Cbx7 recognizes methylated lysine residues on the histone H3 tail and contributes to gene silencing in the context of the Polycomb repressive complex 1 (PRC1). However, our knowledge of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service