Skip to Content
MilliporeSigma

05-1098

Anti-acetyl CoA Carboxylase Antibody, clone 7D2.2

clone 7D2.2, from mouse

Synonym(s):

acetyl-CoA carboxylase 1, acetyl-CoA carboxylase-alpha, acetyl-Coenzyme A carboxylase alpha

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
7D2.2, monoclonal
Application:
WB
Citations:
11
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7D2.2, monoclonal

species reactivity

human, rat

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ACACA(31)
rat ... Acaca(60581)

General description

Acetyl-CoA carboxylase (ACC) is a biotin-dependent enzyme that catalyzes carboxylation of acetyl-CoA to produce malonyl-CoA through its two catalytic activities, biotin carboxylase (BC) and carboxyltransferase (CT). ACC is a multi-subunit enzyme in most prokaryotes, whereas it is a large, multi-domain enzyme in most eukaryotes. The activity of ACC can be controlled at the transcriptional level as well as by small molecule modulators and covalent modification. The human genome contains the genes for two different ACCs - ACACA and ACACB. The activity of the enzyme is controlled by reversible phosphorylation. The activity of the enzyme is inhibited if phosphorylated; the phosphorylation takes place when the hormone glucagon or epinephrine binds to the receptors or the energy status of the cell is low, leading to the activation of the AMP-activated protein kinase. The presence of fatty acid inhibits the activities of the enzyme. When insulin binds to its receptors, it activates a phosphatase to dephosphorylate the enzyme; the activities of the acetyl-CoA carboxylase is thus enhanced. Acetyl-CoA carboxylase has recently become a target in the design of new anti-obesity and antibiotic drugs.
~ 266 kDa Observed

Immunogen

Histidine-tagged recombinant protein corresponding to human acetyl CoA Carboxylase.

Application

Anti-acetyl CoA Carboxylase Antibody, clone 7D2.2 detects level of acetyl CoA Carboxylase & has been published & validated for use in WB.
Research Category
Signaling
Research Sub Category
Insulin/Energy Signaling
Western Blot Analysis: 1 µg/mL from a previous lot detected acetyl CoA Carboxylase on 10 µg of rat heart lysate.

Western Blot (SNAP ID) Analysis: 1 µg/mL from a previous lot detected acetyl CoA Carboxylase on 10 µg of human heart lysate.

Physical form

Format: Purified
Protein G
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human heart lysate
Evaluated by Western Blot in human heart lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected acetyl CoA Carboxylase on 10 µg of human heart lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Replaces: 04-322

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library