Skip to Content
MilliporeSigma

04-745

Anti-trimethyl-Histone H3 (Lys4) Antibody, clone MC315, rabbit monoclonal

culture supernatant, clone MC315, Upstate®

Synonym(s):

H3K4me3, Histone H3 (tri methyl K4)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
MC315, monoclonal
Species reactivity:
human
Application:
ChIP, DB, ICC, IHC, WB, multiplexing
Citations:
299
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

MC315, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

ChIP: suitable (ChIP-seq), dot blot: suitable, immunocytochemistry: suitable, immunohistochemistry: suitable (paraffin), multiplexing: suitable, western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

trimethylation (Lys4)

Gene Information

human ... H3C1(8350)

General description

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure.



The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.
17 kDa

Immunogen

Peptide containing the sequence [RTtrimKQ] in which lysine 4 is trimethylated on human histone H3

Application

Anti-trimethyl-Histone H3 (Lys4) Antibody, clone MC315 is a rabbit monoclonal antibody for detection of trimethyl-Histone H3 (Lys4) also known as H3K4me3, Histone H3 (tri methyl K4) & has been validated in WB, ChIP, DB, Mplex, ChIP-seq.
Immunohistochemistry (Paraffin) Analysis: A 1:250 dilution from a representative lot detected trimethyl-Histone H3 (Lys4) in human colon and human breast cancer tissue sections

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected trimethyl-Histone H3 (Lys4) in HeLa, A431, HUVEC, and NIH3T3 cells.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Biochem/physiol Actions

Histone H3 containing trimethyl-lysine 4 and, to a lesser extent, dimethyl-lysine 4. See COA for specificity data.

Physical form

Cultured supernantant in 0.05% sodium azide.

Preparation Note

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
HeLa acid extract
routinely evaluated by immunoblot on in acid extracted proteins from HeLa cells, but not recombinant unmethylated Histone H3 (Catalog #14-494)

Other Notes

Replaces: 04-791

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Effects of chronic exposure to arsenic and estrogen on epigenetic regulatory genes expression and epigenetic code in human prostate epithelial cells.
Treas, JN; Tyagi, T; Singh, KP
Testing null
Antagonistic roles for BRM and BRG1 SWI/SNF complexes in differentiation.
Flowers, S; Nagl, NG; Beck, GR; Moran, E
The Journal of Biological Chemistry null
Chromatin interaction analysis using paired-end tag sequencing.
Melissa J Fullwood,Yuyuan Han,Chia-Lin Wei,Xiaoan Ruan,Yijun Ruan
Current Protocols in Molecular Biology null



Global Trade Item Number

SKUGTIN
04-74504053252513633