Skip to Content
MilliporeSigma

04-290

Anti-EGFR (CT) Antibody, clone 13G8

clone 13G8, from mouse

Synonym(s):

ERBB, mENA, ERBB1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
13G8, monoclonal
Application:
ELISA, ICC, WB
Citations:
11
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

13G8, monoclonal

species reactivity

mouse, human

manufacturer/tradename

Chemicon®, Upstate®

technique(s)

ELISA: suitable, immunocytochemistry: suitable, western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... EGFR(1956)
mouse ... Egfr(13649)

General description

180 kDa

Immunogen

KLH-conjugated synthetic peptide encompassing the surrounding amino acids of the C terminus in human EGFR

Application

Anti-EGFR (C-terminus) Antibody, clone 13G8 detects level of EGFR (C-terminus) & has been published & validated for use in ELISA, IC & WB.
Research Category
Signaling
Research Sub Category
Growth Factors & Receptors

Biochem/physiol Actions

Recognizes C-terminus domain of EGF receptor (aa1165-1186)

Physical form

100 µg of purified mouse monoclonal IgG lyophilized in PBS containing 0.09% sodium azide, PEG, and sucrose
Format: Purified
Subsequent thiophilic adsorption and size exclusion chromatography

Preparation Note

2 years at -20°C from date of shipment

Analysis Note

Control
Includes EGF treated Hep2G lysate as a positive control
Routinely evaluated by immunoblot.

Other Notes

Replaces: 04-337; 04-338

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library